Skip to Content
Merck
10-Deacetylbaccatin III from Taxus baccata ≥95% (HPLC)

10-Deacetylbaccatin III from Taxus baccata

10-Deacetylbaccatin III from Taxus baccata ≥95% (HPLC)
Empirical Formula (Hill Notation):
C29H36O10
CAS Number:
Molecular Weight:
544.59

Products

Product Category
Brand
Biological Source
Color
Formula Weight
Physical Form
Purity
Activity Spectrum
Mode of Action
Showing 1-1 of 1
Compare
Product Number
Description
Pricing