Solid Formulation: Excipients Used in Pharmaceutical Dosage Forms
![Solid Formulation Excipients Used Pharma Dosage Forms Various pink and purple pills and capsules scattered on a bright yellow background. One prominent round pink pill in the middle of the picture features the letter "M" embossed on it.](/deepweb/assets/sigmaaldrich/marketing/global/images/categories/pharma-and-biopharma-manufacturing/solid-formulation-excipients-pharma-dosage-forms/solid-formulation-excipients-pharma-dosage-forms.jpg)
Solid dose formulations are one of the most established dosage forms in pharmaceuticals, and yet the path to market can still be blocked by critical obstacles such as active pharmaceutical ingredient (API) stability, release kinetics, and bioavailability limitations.
Our high-quality pharmaceutical raw materials and functionalized excipients form a comprehensive portfolio that addresses all your most pressing challenges in solid formulation manufacturing, with products that include:
Read more about
Solid Formulation Products
PARTECK® FUNCTIONAL EXCIPIENTS: FOR YOUR SOLID FORMULATION DEVELOPMENT
When you design a solid dosage formulation, there are many factors to consider: the active pharmaceutical ingredient (API), bioavailability enhancement, release kinetics, specific dosage, packaging, shelf life, and more. Excipients are the backbone of a formulation and affect final formulation properties such as processability, stability and performance. Therefore, it is important to identify an excipient that has the correct functionality and is of high quality.
Our Parteck® functional excipients were designed with these formulator’s challenges in mind and have additional functionalities beyond standard excipients. These excipients are particle optimized and feature unique particle properties tailored for tableting processes, for specific drug delivery technologies, or for solubility enhancement. Thus, our Parteck® functional excipients ensure excellent performance in formulation development and manufacturing. Backed by the Emprove® Program with its stringent quality control, as well as regulatory support and documentation, Parteck® excipients help to speed up your approval processes.
A comprehensive range of functional excipients, facilitating high tablet hardness at low compression forces, short disintegration times as well as high and low API dosages, targeting:
- Direct compression
- Granulation
- Tablet coatings
Particle-optimized excipients targeted at different administration routes with the aim to support your desired release kinetics in:
- Oral sustained release formulations
- Orally disintegrating tablets (ODT)
- Dry powder inhalation
Innovative excipients supporting a variety of solubility enhancement approaches to improve API solubility through formulation including:
- Hot melt extrusion
- Spray drying
- Dissolution enhancers
- Inorganic drug carriers
Specialized excipients for additive manufacturing targeting specifically the pharmaceutical industry with a focus on melt-based 3D printing technologies such as:
- Melt-drop deposition
Portfolio of excipients optimized for excellent performance in Continuous Manufacturing:
- Parteck® LM self-lubricating filler/binder
- Parteck® excipient range designed for direct compression and dry granulation
- Parteck® M mannitol suitable for twin screw granulation
Platform technology based on mesoporous silica to improve formulation efficiency and de-risk solid state variation during drug development through:
- Stabilization of unstable polymorphs
- Optimization of API solid-state properties
In addition, you can rely on our industry-leading Emprove® Program to meet today’s complex regulatory challenges with a superior combination of comprehensive documentation and transparency. Our high-quality raw materials, regulatory expertise, and dedicated support provide everything you need to simplify supplier qualification, speed up processes, and reduce your total cost of ownership.
Related Product Resources
- Article: API Solubility and Dissolution Enhancement Via Formulation
Our overview of API solubility and dissolution enhancement methods can help you decide between the many options to take during formulation.
- Article: Solid Formulation Development Using Melt-based 3D Printing Technologies
3D printing can overcome challenges during formulation development, with a focus on enhancing API bioavailability in solid dispersions. Find out how in this technical article.
- Article: Improving API Solubility Using Hot Melt Extrusion Formulation With Polyvinyl Alcohol
Find out how Hot Melt Extrusion with polyvinyl alcohol (PVA) is used to enhance API solubility.
- Download: Formulation Handbook
In this formulation handbook, we have compiled information and formulation examples that can help you when developing your formulation.
Web Tools
- Formulation Product Finder
Quickly sort our excipients and API portfolio by dosage form, application, and many other parameters.
- Pharmaceutical Application Guide
Explore products and services for each step of your pharmaceutical manufacturing process.
- Risk Mitigation Tool
Get guidance through the challenges and quality requirements of your bio-manufacturing process.