SY3H94E1CB36
Synthonix Corporation
4-(Ethoxycarbonyl)phenylzinc bromide, 0.50 M in THF
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
bromo[4-(ethoxycarbonyl)phenyl]zinc
Inchi Code
1S/C9H9O2.BrH.Zn/c1-2-11-9(10)8-6-4-3-5-7-8;;/h4-7H,2H2,1H3;1H;/q;;+1/p-1
InChI key
CYJKOKZYUACUHZ-UHFFFAOYSA-M
storage temp.
Refrigerator
Purity
0.50
Country of Origin
US
Shipping Temperature
Room Temperature
Physical Form
Solution
Product Link
https://synthonix.com/73216.html
Signal Word
Danger
Hazard Statements
MSDS
https://synthonix.com/73216.html
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service