Skip to Content
Merck
All Photos(1)

Key Documents

74013

Supelco

4-Methyl-2-oxovaleric acid-13C6 sodium salt

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13C6H9NaO3
CAS Number:
Molecular Weight:
158.08
MDL number:
UNSPSC Code:
12352106
NACRES:
NA.24

grade

analytical standard

Assay

≥90.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

storage temp.

2-8°C

SMILES string

[13CH3][13CH]([13CH3])[13CH2][13C]([13C](O[Na])=O)=O

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service