Key Documents
58284
Screen for Valco® Pre-Column Filter
pore size 2.0 μm, stainless steel
About This Item
UNSPSC Code:
41115700
Pricing and availability is not currently available.
material
stainless steel
packaging
pkg of 10 ea
pore size
2.0 μm
Recommended products
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Legal Information
Valco is a registered trademark of Valco Instruments Co. Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Articles
Correct common HPLC issues to restore column performance, addressing peak problems, retention time, noise, and resolution with chromatograms.