Skip to Content
Merck
All Photos(1)

Documents

47577-U

Supelco

EPA GRO Mix

certified reference material, in methanol (varied)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Agency

EPA
610

product line

TraceCERT®

CofA

current certificate can be downloaded

feature

standard type calibration

packaging

ampule of 1 mL

concentration

in methanol (varied)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

suitability

suitable for GRO (Fuels (TPH))

application(s)

environmental

format

multi-component solution

storage temp.

2-30°C

General description

EPA GRO Mix is a multi-component gasoline range organics (GRO) standard containing certified reference materials (CRMs) such as benzene, ethylbenzene, heptane, 2-methylpentane, toluene, 1,2,4-trimethylbenzene, 2,2,4-trimethylpentane, o-xylene, and m-xylene. The GRO-like methods are used to quantify volatile analytes and require purge and trap or direct injection.

Application

EPA GRO Mix can be used as a certified reference material (CRM) standard to quantify the volatile organic impurities in inorganic coagulants used in drinking water and human breath samples by gas chromatography-mass spectrometry (GC-MS).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Analyte

Description

    Benzene 500 μg/mL
    Ethylbenzene 500 μg/mL
    Heptane 500 μg/mL
    2-Methylpentane 1500 μg/mL
    Toluene 1500 μg/mL
    1,2,4-Trimethylbenzene 1000 μg/mL
    2,2,4-Trimethylpentane 1500 μg/mL
    o-Xylene 1000 μg/mL
    m-Xylene 1000 μg/mL
    See All (9)

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Screening analysis of volatile organic contaminants in commercial inorganic coagulants used for drinking water treatment
Petri M, et al.
Journal of Environmental Management, 91(1), 142-148 (2009)
Stephen J Fowler et al.
Thorax, 70(4), 320-325 (2015-02-11)
Healthcare associated infections, including ventilator associated pneumonia, are difficult to diagnose and treat, and are associated with significant morbidity, mortality and cost. We aimed to demonstrate proof of concept that breath volatile profiles were associated with the presence of clinically

Protocols

US EPA Method 8015 (modified): GC Analysis of Gasoline Range Organics (GRO) on SLB®-5ms, Direct Injection

There are three types of analyses: GRO, DRO, and TPH. The published methods are simply variations of these to meet the different analytical needs of the individual agencies.

Chromatograms

suitable for GC

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service