Skip to Content
Merck
All Photos(1)

Documents

24938

Supelco

Purge/Trap I Vocarb® 4000

for use with OI Analytical Eclipse 4660, 4560

Synonym(s):

PURGE TRAP I, O.I. ANALYTICAL 4560

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700

Agency

suitable for EPA 601

application(s)

environmental

compatibility

for use with OI Analytical Eclipse 4660, 4560

Looking for similar products? Visit Product Comparison Guide

General description

Tubes used in purge and trap analyses generally are packed with multiple beds of adsorbent materials, so that a broad range of polar and nonpolar, high and low molecular weight compounds can be trapped in a single tube. Each bed protects the next, increasingly active bed, by preventing compounds from being held so strongly that they cannot be desorbed quickly without decomposition. During the purge phase of sampling, lower molecular weight compounds pass through the initial adsorbent beds but are trapped by succeeding beds. During desorption, the carrier gas passes through the trap in the reverse direction of purge flow, so that higher molecular weight compounds never come in contact with the stronger (innermost) adsorbents.

Application

Purge/Trap I Vocarb® 4000 may be used as a purge and trap system in the gas chromatographic analysis of volatile organic compounds.

Legal Information

Vocarb is a registered trademark of Siemens Industry, Inc.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reactions of vinyl chloride and methanol in a quadrupole ion trap mass spectrometer during VOC analysis.
Bian L, et al.
Environmental Science & Technology, 33(9), 1528-1533 (1999)

Related Content

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service