Skip to Content
Merck
All Photos(1)

Key Documents

Z688061

E-Vac aspiration waste system

with 4 L polypropylene bottle, barbed tube fittings lid

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.22

manufacturer/tradename

Cole Parmer 1305031

Looking for similar products? Visit Product Comparison Guide

General description

Safe, self-contained waste system provides an alternative to traditional in-house vacuum!

These compact waste systems are ideal for use with critical or dangerous liquids or supernatants, pathogens, or any class III or IV biohazard laboratories. Quiet, oil-free membrane pump has a range from –250 mm to 650 mm Hg. Once target vacuum is reached, pump automatically switches off. Pump automatically switches on when vacuum pressure is applied allowing it to maintain a constant vacuum pressure—perfect for gentle aspiration of small plates and dishes, as well as rapid emptying of larger containers.

Easy-to-clean stainless steel housing is UV-resistant for use in laminar hoods.

The E-Vac bottles can be autoclaved for 20 minutes at 121°C to prevent laboratory contamination. Base units are illuminated to allow visual inspection of waste levels in the bottle. Dual hydrophobic filters prevent liquids from reaching the pump housing and contaminating the system.

E-Vac is available with a 4-L polypropylene bottle or a 3-L safety glass bottle. Polypropylene and glass bottles offer lids equipped with quick-release tubing connectors for safe and convenient tubing connections; and level detection sensor that automatically shuts the pump off when the bottle is full. Polypropylene bottle also available with lid fitted with standard barbed tube fittings.

  • Compact, user-friendly design
  • Level detection system prevents bottle overfill
  • Control knob for infinite vacuum setting from -250 mbar to -650 mbar
  • Includes: HandE-Vac hand operator with single-channel plastic adapter
  • Includes: All wetted parts such as bottle, lid, tubing, connectors and hand operator are autoclavable

The E-Vac is CE compliant. The supplied international mains plug (100-240V 50/60Hz) is also CE compliant. It includes adaptors for the US, UK, Australia and a two prong European.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service