Skip to Content
Merck
All Photos(4)

Key Documents

WH0006297M10

Sigma-Aldrich

Monoclonal Anti-SALL2 antibody produced in mouse

clone 3F7, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-HSAL2, Anti-KIAA0360, Anti-p150(Sal2), Anti-sal-like 2 (Drosophila)

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
R$3,068.00

R$3,068.00


Estimated to ship onApril 25, 2025



Select a Size

Change View
100 μG
R$3,068.00

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

R$3,068.00


Estimated to ship onApril 25, 2025


biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

3F7, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG2aκ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SALL2(6297)

Related Categories

General description

SALL2 (sal-like 2, Drosophila) is a multi-zinc finger transcription factor, belongs to the Drosophila homeotic Spalt-like family of developmental transcription factor genes. It is expressed during development of human retina at the time of optic fissure closure.

Immunogen

SALL2 (AAH24245, 1 a.a. ~ 198 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
MAHESERSSRLGVPCGEPAELGGDASEEDHPQVCAKCCAQFTDPTEFLAHQNACSTDPPVMVIIGGQENPNNSSASSEPRPEGHNNPQVMDTEHSNPPDSGSSVPTDPTWGPERRGEESPGHFLVAATEPVCGIPVKWPAHEALEFQLHLHYHSKPGPTSAVWPRNCGWEGASNNGIQGSQGEDSPPPISASCTQGSA

Biochem/physiol Actions

SALL2 majorly is associated with the growth arrest and pro-apoptotic functions. It is involved in eye morphogenesis. Alteration in the gene function causes ocular coloboma in humans and mice. It has ability to supress the c-MYC transcription by binding to the nuclease hypersensitive element of the c-MYC promoter.

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daniel Kelberman et al.
Human molecular genetics, 23(10), 2511-2526 (2014-01-15)
Ocular coloboma is a congenital defect resulting from failure of normal closure of the optic fissure during embryonic eye development. This birth defect causes childhood blindness worldwide, yet the genetic etiology is poorly understood. Here, we identified a novel homozygous
Hongcang Gu et al.
Biochimica et biophysica acta, 1809(4-6), 276-283 (2011-03-03)
The product of the SALL2 protein p150(Sal2) is a multi-zinc finger transcription factor with growth arrest and proapoptotic functions that overlap those of p53. Its DNA-binding properties are unknown. We have used a modified SELEX procedure with purified p150(Sal2) and
Chang Kyoo Sung et al.
PloS one, 7(9), e46486-e46486 (2012-10-03)
p150, product of the SALL2 gene, is a binding partner of the polyoma virus large T antigen and a putative tumor suppressor. p150 binds to the nuclease hypersensitive element of the c-MYC promoter and represses c-MYC transcription. Overexpression of p150

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service