Skip to Content
Merck
All Photos(2)

Key Documents

WH0004857M7

Sigma-Aldrich

Monoclonal Anti-NOVA1 antibody produced in mouse

clone 3F3, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-Nova1, Anti-neuro-oncological ventral antigen 1

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

3F3, monoclonal

form

buffered aqueous solution

species reactivity

human, rat, mouse

technique(s)

indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG2aκ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... NOVA1(4857)

General description

Neuro-oncological ventral antigen 1 (NOVA1) is expressed in fibroblasts. It is part of the Nova family of neuron-specific RNA-binding proteins. The NOVA1 gene is localized on human chromosome 14q12.[1][2][3]

Immunogen

NOVA1 (NP_002506, 408 a.a. ~ 507 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
SAILGTEKSTDGSKDVVEIAVPENLVGAILGKGGKTLVEYQELTGARIQISKKGEFVPGTRNRKVTITGTPAATQAAQYLITQRITYEQGVRAANPQKVG

Biochem/physiol Actions

NOVA1(Neuro-oncological ventral antigen 1) is associated with several post-transcriptional regulation of RNA metabolism including RNA splicing, editing to transport, localization, and degradation.[4] It may be involved in mediating neuronal responsiveness.[4] It encodes a protein which has homology with the RNA-binding protein hnRNP K, the yeast splicing protein MER1. Since, it has some homology with hnRNP K, it may influence the regulation of RNA splicing or metabolism in developing neurons.[5] The importance of NOVA1 as prognostic marker has been reported in HCC (Hepatocellular carcinoma).[6] Alteration in gene causes a disorder associated with breast cancer and motor dysfunction i.e. paraneoplastic opsoclonus myoclonus ataxia (POMA).[7]

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

14q12 and severe Rett-like phenotypes: new clinical insights and physical mapping of FOXG1-regulatory elements
Lila Allou
European Journal of Human Genetics (2012)
Implications of NOVA1 suppression within the microenvironment of gastric cancer: association with immune cell dysregulation.
Kim EK
Gastric cancer : official journal of the International Gastric Cancer Association and the Japanese Gastric Cancer Association (2017)
MiR-181b-5p downregulates NOVA1 to suppress proliferation, migration and invasion and promote apoptosis in astrocytoma.
Zhi F
PLoS ONE (2014)
Mylène Hervé et al.
Disease models & mechanisms, 9(8), 899-909 (2016-08-03)
Familial dysautonomia (FD) is a rare neurodegenerative disease caused by a mutation in intron 20 of the IKBKAP gene (c.2204+6T>C), leading to tissue-specific skipping of exon 20 and a decrease in the synthesis of the encoded protein IKAP (also known
R J Buckanovich et al.
Neuron, 11(4), 657-672 (1993-10-01)
Paraneoplastic opsoclonus-ataxia, a disorder of motor control, develops in breast or lung cancer patients who harbor an antibody (Ri) that recognizes their tumors and a nuclear neuronal protein of 55 kd. We have characterized a gene, Nova, encoding an antigen

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service