Skip to Content
Merck
All Photos(1)

Key Documents

T2007

Sigma-Aldrich

Tyr-Tyr-Tyr

Synonym(s):

L-tyrosyl-L-tyrosyl- L-tyrosine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H29N3O7
CAS Number:
Molecular Weight:
507.54
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.26

Assay

≥98% (TLC)

Quality Level

form

powder

technique(s)

HPLC: suitable

color

white

storage temp.

−20°C

SMILES string

NC(Cc1ccc(O)cc1)C(=O)NC(Cc2ccc(O)cc2)C(=O)NC(Cc3ccc(O)cc3)C(O)=O

InChI

1S/C27H29N3O7/c28-22(13-16-1-7-19(31)8-2-16)25(34)29-23(14-17-3-9-20(32)10-4-17)26(35)30-24(27(36)37)15-18-5-11-21(33)12-6-18/h1-12,22-24,31-33H,13-15,28H2,(H,29,34)(H,30,35)(H,36,37)

InChI key

RMRFSFXLFWWAJZ-UHFFFAOYSA-N

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Asako Narai-Kanayama et al.
Journal of biotechnology, 150(3), 343-347 (2010-09-28)
Papain-catalyzed polymerization of L-tyrosine ethyl ester in aqueous media was efficient for synthesis of oligo-tyrosine peptides having angiotensin I-converting enzyme inhibitory activity. Di-, tri-, and tetra-tyrosine accumulated in the soluble fraction of reaction mixture. On the other hand, the peptide
Anne Beilvert et al.
Chemical communications (Cambridge, England), 47(19), 5506-5508 (2011-04-02)
This communication reports the synthesis, characterization and in vivo evaluation in mice of a new tri-tyrosine conjugated MR contrast agent, which may help to identify vulnerable plaques in atherosclerosis by targeting the lipid core.
Javiera Cortés-Ríos et al.
Analytical biochemistry, 608, 113904-113904 (2020-08-18)
Amongst the available methodologies for protein determination, the bicinchoninic acid (BCA) assay highlights for its simplicity, sensitivity, repeatability and reproducibility. Nevertheless, in spite that the general principle behind this methodology is known, there are still unanswered questions regarding the chemistry
Shun Hirota et al.
Biochemical and biophysical research communications, 314(2), 452-458 (2004-01-22)
Effect of a hydrophobic peptide on folding of oxidized cytochrome c (cyt c) is studied with trityrosine. Folding of cyt c was initiated by pH jump from 2.3 (acid-unfolded) to 4.2 (folded). The Soret band of the 2-ms transient absorption

Articles

DISCOVER Bioactive Small Molecules for Neuroscience

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service