Skip to Content
Merck
All Photos(1)

Documents

SRP5194

Sigma-Aldrich

I?Bα, GST tagged human

recombinant, expressed in E. coli, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

MAD-3, NFKBI, NFKBIA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

recombinant

expressed in E. coli

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

mol wt

~61 kDa

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... NFKBIA(4792)

General description

IκBα is an inhibitor of the NFκB complex and inactivates the NFκB by trapping it in the cytoplasm. Phosphorylation of serine residues on the IκB protein by IκB kinases (IKKs) marks it for destruction via the ubiquitination pathway, thereby allowing the activation of the NFκB complex. Synthetic glucocorticoid such as dexamethasone display anti-inflammatory effects by inducing the increased synthesis of the IκB protein thereby inhibiting the activity of the NFκB complex. Overexpression of the IκBα gene in fibroblasts leads to inhibition of production of IL-6, TNF receptor, MMP-1, MMP-3 and MMP-13.

Physical form

Supplied in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol.

Preparation Note

after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N Auphan et al.
Science (New York, N.Y.), 270(5234), 286-290 (1995-10-13)
Glucocorticoids are among the most potent anti-inflammatory and immunosuppressive agents. They inhibit synthesis of almost all known cytokines and of several cell surface molecules required for immune function, but the mechanism underlying this activity has been unclear. Here it is
Jan Bondeson et al.
The Journal of rheumatology, 34(3), 523-533 (2007-02-14)
To investigate the role of the transcription factor nuclear factor-kB (NF-kappaB) in promoting inflammatory and destructive responses in human osteoarthritis (OA) synovial fibroblasts, by assessing the effect of NF-kappaB blockade on the production of cytokines and destructive enzymes. Infection with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service