Skip to Content
Merck
All Photos(1)

Documents

SRP0707

Sigma-Aldrich

PAD Cocktail, Active

recombinant

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.77

recombinant

expressed in baculovirus infected Sf9 cells

Assay

70-95% (SDS-PAGE)

form

frozen liquid

mol wt

~95-100 kDa

concentration

0.5 μg/μL (0.1 ug/uL each PAD)

accession no.

BC025718
BC049947
NM_013358
NM_016233
NM_207421

shipped in

dry ice

storage temp.

−70°C

Application

Useful for citrullination and Western blot.

Biochem/physiol Actions

The five full length recombinant PAD proteins (1, 2, 3, 4 and 6) expressed by baculovirus in Sf9 insect cells using N-terminal GST tags. This mixture is ideally suited for in vitro citrullination of protein targets.

Reconstitution

Formulated in 50mM Tris-HCl, pH 7.5, 50mM NaCl, 10mM glutathione, 0.25 mM DTT, 0.1 mM PMSF, with 25% glycerol.

Storage and Stability

Store in aliquots at -70°C. Avoid freeze/thaw cycles.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service