Skip to Content
Merck
All Photos(1)

Documents

SML2810

Sigma-Aldrich

Prostetin Trifluoroacetic acid salt

≥98% (HPLC)

Synonym(s):

2-(4-(4-(3-(3-Chlorophenyl)-1H-pyrrolo[2,3-b]pyridin-5-yl)benzyl)piperazin-1-yl)ethan-1-ol Trifluoroacetic acid salt, 2-(4-(4-(3-(3-Chlorophenyl)-1H-pyrrolo[2,3-b]pyridin-5-yl)benzyl)piperazin-1-yl)ethanol Trifluoroacetic acid salt, 4-[[4-[3-(3-Chlorophenyl)-1H-pyrrolo[2,3-b]pyridin-5-yl]phenyl]methyl]-1-piperazineethanol Trifluoroacetic acid salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H27ClN4O · xC2HF3O2
Molecular Weight:
446.97 (free base basis)
UNSPSC Code:
12352106
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

Biochem/physiol Actions

Prostetin is an orally available, blood-brain barrier-penetrant, potent and metabolically stable inhibitor of the mitogen-activated protein kinase kinase kinase kinase (MAP4K, or HGK) that exhibits neuroprotective and anti-inflammatory properties. Prostetin is more potent than URMC-099 in motor neuron survival assay. Prostetin rescues CPA-induced neurodegeneration in motor neurons.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pieter H Bos et al.
Cell chemical biology, 26(12), 1703-1715 (2019-11-05)
Disease-causing mutations in many neurodegenerative disorders lead to proteinopathies that trigger endoplasmic reticulum (ER) stress. However, few therapeutic options exist for patients with these diseases. Using an in vitro screening platform to identify compounds that protect human motor neurons from ER

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service