Skip to Content
Merck
All Photos(1)

Key Documents

SMB00903

Sigma-Aldrich

Rebaudioside F

Synonym(s):

Rebaudioside F

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C43H68O22
Molecular Weight:
936.99
MDL number:
UNSPSC Code:
12352205

biological source

Stevia sp.

Assay

≥95% (HPLC)

form

solid

color

white to off-white

shipped in

wet ice

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Other Notes

Rebaudioside F is a diterpenoid glycoside isolated from the leaves of a high rebausioside C producing line of Stevia rebaudiana.
To gain a comprehensive understanding of our extensive range of Oligosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service