Skip to Content
Merck
All Photos(1)

Key Documents

SAB4701038

Sigma-Aldrich

Monoclonal Anti-Actin (muscle) antibody produced in mouse

clone HHF35

Synonym(s):

Muscle specific actin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

HHF35, monoclonal

form

buffered aqueous solution

species reactivity

rabbit, nonhuman primates, cat, rat, dog

concentration

1 mg/mL

technique(s)

western blot: 1 μg/mL

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

General description

The mouse monoclonal antibody HHF35 recognizes muscle-specific alpha and gamma actin (42 kDa) in various species. This antibody stains skeletal, smooth and myocardial cells as well as myoepithelial cells and pericytes of small vessels. It is a widely used marker of muscle and muscle-derived cells.

Immunogen

SDS extracted protein fraction of human myocardium

Physical form

Solution in phosphate buffered saline with 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service