Skip to Content
Merck
All Photos(9)

Documents

RNB100

Sigma-Aldrich

GenElute Total RNA Purification Kit

greener alternative

Synonym(s):

Total RNA Extraction Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105600

feature

DNA free (removal without the use of enzyme)

storage condition

room temperature

greener alternative product characteristics

Designing Safer Chemicals
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

technique(s)

RT-PCR: suitable

greener alternative category

shipped in

ambient

storage temp.

room temp

General description

Sigma′s GenElute Total RNA Purification Kit provides a rapid method for the isolation and purification of total RNA from cultured animal cells, tissue samples, blood, plasma, serum, bacteria, yeast, fungi, plants and viruses. The kit purifies all sizes of RNA, from large mRNA and ribosomal RNA down to microRNA (miRNA) and small interfering RNA (siRNA). The RNA is preferentially purified from other cellular components such as proteins, without the use of phenol or chloroform. The purified RNA is of the highest integrity, and can be used in a number of downstream applications including real time PCR, reverse transcription PCR, Northern blotting, RNase protection and primer extension, and expression array assays.

  • Isolate true total RNA including siRNA and microRNA
  • Rapidly remove contaminating genomic DNA without the use of enzymes
  • No phenol or chloroform extractions
  • Purify high-quality RNA in 20 minutes
  • Extract RNA from as little as a single cell
  • Isolate from a wide variety of specimens

Application

GenElute Total RNA Purification Kit has been used to:
  • Quantitative, real-time RT-PCR for both large mRNA and small RNA including miRNA
  • RT-PCR for both large mRNA and small RNA including miRNA
  • Expression profiling
  • Next Generation Sequencing for RNA and miRNA
  • miRNA from plasma for discovery
  • microRNA cloning and amplification
  • PCR-based virus detection
  • PCR-based viable bacteria detection
  • Northern blotting
  • RNase protection
  • Primer extension
  • miRNA array
  • RNA-Seq
  • Nanostring
  • Fluidigm
  • Droplet/Digital PCR
  • Isolate total RNA of rat kidney tissues
GenElute Total RNA Purification Kit is used for the extraction of total RNA:
  • From whole blood samples in the detection of Hepatitis C Virus using a reverse transcriptase-polymerase spiral reaction (RT-PSR) system.
  • From an overnight culture of bacterial strains in TSB medium in determining the effects of curcumin and silymarin on virulence and carbapenemase genes among multidrug-resistant Escherichia coli clinical isolates.

Features and Benefits

  • No phenol: chloroform extractions - Total RNA is isolated without the use of harmful chemicals such as phenol or chloroform
  • Isolate total RNA from very small samples - Total RNA has been isolated and detected from as little as a single animal cell
  • Extremely efficient isolation of low abundance microRNA - GenElute′s Total RNA Purification Kit has been shown to be extremely efficient at recovering low abundance microRNA from plasma samples
  • Isolate a diversity of RNA species - All RNA species can be isolated, from large mRNA and ribosomal RNA down to microRNA (miRNA) and small interfering RNA (siRNA)
  • Fast and easy processing - Rapid spin-column format allows for the processing of 10 samples in 20 minutes
  • Isolate total RNA from a broad input source - Total RNA has been isolated from cultured animal cells, small tissue samples, LCM samples, bacteria, yeast, fungi, plants, viruses and various bodily fluid including blood, plasma, serum, saliva, nasal or throat swabs
  • No need for carrier RNA - Isolate all sizes of RNA without the use of carrier RNA
  • Multiple kit sizes available - This kit is available in both 50 prep and 100 prep sizes

Unit Definition

Preps

Other Notes

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has Inherently Safer Chemistry, compared to the standard use of phenol and chloroform to perform DNA extractions.

Legal Information

GenElute is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rapid and Sensitive Detection of Hepatitis C Virus in Clinical Blood Samples Using Reverse Transcriptase Polymerase Spiral Reaction
Sun W, et al.
Journal of microbiology and biotechnology, 30(3), 459-468 (2020)
Valentina Lagos-Rodríguez et al.
Pain, 161(12), 2786-2797 (2020-07-14)
Glial reactivity in the dorsal horn of the spinal cord is a hallmark in most chronic pain conditions. Neuroinflammation-associated reactive glia, in particular astrocytes, have been shown to exhibit reduced mitochondrial respiratory function. Here, we studied the mitochondrial function at
Jing-Zhuang Zhao et al.
Journal of virological methods, 265, 84-90 (2019-01-08)
Reverse genetics systems are powerful tools for understanding the virulence mechanisms and gene functions of negative-sense RNA viruses. The reverse genetics systems commonly used for recombinant infectious hematopoietic necrosis virus (IHNV) are based on vaccinia virus infection. To avoid the
Wenying Sun et al.
Journal of microbiology and biotechnology, 30(3), 459-468 (2020-01-03)
This study established a new polymerase spiral reaction (PSR) that combines with reverse transcription reactions for HCV detection targeting 5'UTR gene. To avoid cross-contamination of aerosols, an isothermal amplification tube (IAT), as a separate containment control, was used to judge
Chunjie Zhang et al.
Oncology letters, 21(3), 210-210 (2021-02-09)
Long non-coding RNAs (lncRNAs) play an important role in gene regulation. Several lncRNAs have been demonstrated to be associated with the diagnosis and prognosis of non-small cell lung cancer (NSCLC). The present study aimed to investigate the role of lncRNA

Protocols

Instructions for purifying viral RNA using GenElute™ RNA purification spin prep kits.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service