Skip to Content
Merck
All Photos(4)

Key Documents

P7627

Sigma-Aldrich

6-Phosphogluconic acid barium salt hydrate

Grade VI, ≥90%, powder

Synonym(s):

6-PG-Ba, D-Gluconate 6-phosphate barium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H10Ba1.5O10P · xH2O
CAS Number:
Molecular Weight:
479.10 (anhydrous basis)
Beilstein:
1729373
EC Number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

biological source

synthetic (organic)

type

Grade VI

Assay

≥90%

form

powder

color

white

storage temp.

−20°C

SMILES string

[O-]P(OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C([O-])=O)([O-])=O.[H]O[H].C

InChI

1S/C6H13O10P.CH4.H2O/c7-2(1-16-17(13,14)15)3(8)4(9)5(10)6(11)12;;/h2-5,7-10H,1H2,(H,11,12)(H2,13,14,15);1H4;1H2/p-3/t2-,3-,4+,5-;;/m1../s1

InChI key

VINGHIAQWNFZIR-ZBHRUSISSA-K

Looking for similar products? Visit Product Comparison Guide

General description

suitable for glucose-6-phosphate dehydrogenase tests

Application

6-Phosphogluconic acid has been used in a study to assess the biochemistry of Coxiella burnetti. It has also been used in a study to investigate the enzymatic transfer of hydrogen.

Preparation Note

Prepared by a modification of the procedure of Seegmiller, J.E., et al., J. Biol. Chem., 192, 175 (1951).

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hiroyoshi Matsumura et al.
Journal of molecular biology, 422(1), 75-86 (2012-05-23)
The key enzyme of plant photosynthesis, D-ribulose 1,5-bisphosphate carboxylase/oxygenase (Rubisco), must be activated to become catalytically competent via the carbamylation of Lys201 of the large subunit and subsequent stabilization by Mg(2+) coordination. Many biochemical studies have reported that reduced nicotinamide
Teresa del Castillo et al.
Journal of bacteriology, 189(14), 5142-5152 (2007-05-08)
In this study, we show that glucose catabolism in Pseudomonas putida occurs through the simultaneous operation of three pathways that converge at the level of 6-phosphogluconate, which is metabolized by the Edd and Eda Entner/Doudoroff enzymes to central metabolites. When
Paolo A Calligari et al.
Proteins, 80(4), 1196-1210 (2012-01-26)
Nuclear magnetic resonance is used to investigate the backbone dynamics in 6-phosphogluconolactonase from Trypanosoma brucei (Tb6PGL) with (holo-) and without (apo-) 6-phosphogluconic acid as ligand. Relaxation data were analyzed using the model-free approach and reduced spectral density mapping. Comparison of
Hulya Ayar-Kayali
Preparative biochemistry & biotechnology, 41(1), 94-105 (2011-01-14)
In vivo pentose phosphate pathway (PPP) enzymes such as glucose-6-phosphate dehydrogenase (G6PDH), 6-phosphogluconate dehydrogenase (6PGDH), and transaldolase (TAL) activities as well as ATP- and ADP-level variations of Amycolatopsis orientalis were investigated with respect to glucose concentration and incubation period. G6PDH
Luísa Ramos et al.
Carbohydrate research, 339(13), 2225-2232 (2004-09-01)
Multinuclear ((1)H, (13)C, (17)O, (31)P, (95)Mo, (183)W) magnetic resonance spectroscopy (1D and 2D) has been used to show that 6-phospho-d-gluconic acid forms three complexes with tungsten(VI) and six complexes with molybdenum(VI) in aqueous solution, depending on pH and concentration. Two

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service