Skip to Content
Merck
All Photos(1)

Documents

P0417

Sigma-Aldrich

Pentadecylamine

~97% (capillary GC)

Synonym(s):

1-Aminopentadecane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)14NH2
CAS Number:
Molecular Weight:
227.43
Beilstein:
1744557
EC Number:
MDL number:
UNSPSC Code:
12352200

vapor pressure

1 mmHg ( 120.5 °C)

Assay

~97% (capillary GC)

bp

299-301 °C (lit.)

mp

35-37 °C (lit.)

storage temp.

2-8°C

SMILES string

CCCCCCCCCCCCCCCN

InChI

1S/C15H33N/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16/h2-16H2,1H3

InChI key

JPZYXGPCHFZBHO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service