Skip to Content
Merck
All Photos(1)

Key Documents

N9398

Sigma-Aldrich

N-Acetyl-[Leu28, Leu31]-Neuropeptide Y Fragment 24-36

≥98% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

Product Name

N-Acetyl-[Leu28, Leu31]-Neuropeptide Y Fragment 24-36, ≥98% (HPLC), powder

Assay

≥98% (HPLC)

form

powder

UniProt accession no.

storage temp.

−20°C

SMILES string

CC(C)C[C@H](NC(C)=O)C(=O)N[C@@H](CCCNC(N)=N)C(=O)N[C@@H](Cc1c[nH]cn1)C(=O)N[C@@H](Cc2ccc(O)cc2)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](CC(N)=O)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H]([C@@H](C)O)C(=O)N[C@@H](CCCNC(N)=N)C(=O)N[C@@H](CCC(N)=O)C(=O)N[C@@H](CCCNC(N)=N)C(=O)N[C@@H](Cc3ccc(O)cc3)C(N)=O

InChI

1S/C81H131N27O19/c1-40(2)30-56(96-45(10)110)71(120)98-52(15-12-28-93-80(87)88)69(118)106-61(36-48-38-91-39-95-48)75(124)105-60(35-47-19-23-50(112)24-20-47)74(123)102-58(32-42(5)6)73(122)107-62(37-64(83)114)76(125)103-57(31-41(3)4)72(121)104-59(33-43(7)8)77(126)108-65(44(9)109)78(127)100-53(16-13-29-94-81(89)90)67(116)99-54(25-26-63(82)113)70(119)97-51(14-11-27-92-79(85)86)68(117)101-55(66(84)115)34-46-17-21-49(111)22-18-46/h17-24,38-44,51-62,65,109,111-112H,11-16,25-37H2,1-10H3,(H2,82,113)(H2,83,114)(H2,84,115)(H,91,95)(H,96,110)(H,97,119)(H,98,120)(H,99,116)(H,100,127)(H,101,117)(H,102,123)(H,103,125)(H,104,121)(H,105,124)(H,106,118)(H,107,122)(H,108,126)(H4,85,86,92)(H4,87,88,93)(H4,89,90,94)/t44-,51+,52+,53+,54+,55+,56+,57+,58+,59+,60+,61+,62+,65+/m1/s1

InChI key

LZKBNKKHNKAGAE-ZQWBWPFKSA-N

Gene Information

human ... NPY(4852)

Amino Acid Sequence

Ac-Leu-Arg-His-Tyr-Leu-Asn-Leu-Leu-Thr-Arg-Gln-Arg-Tyr-NH2

Biochem/physiol Actions

N-Acetyl-[Leu28, Leu31]-Neuropeptide Y Fragment 24-36 is a neuropeptide Y-Y2 receptor agonist.
Selective neuropeptide Y-Y2 receptor agonist.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ernie Yulyaningsih et al.
British journal of pharmacology, 163(6), 1170-1202 (2011-05-07)
The neuropeptide Y system has proven to be one of the most important regulators of feeding behaviour and energy homeostasis, thus presenting great potential as a therapeutic target for the treatment of disorders such as obesity and at the other
D J Tracey et al.
Brain research, 669(2), 245-254 (1995-01-16)
Injury of peripheral nerves often results in hyperalgesia (an increased sensitivity to painful stimuli). This hyperalgesia is mediated in part by sympathetic neurotransmitters. We examined the effect of neuropeptide Y (NPY), specific Y1 and Y2 agonists, and an NPY antagonist
E K Potter et al.
European journal of pharmacology, 267(3), 253-262 (1994-05-17)
We have carried out functional and in vitro studies on a novel analog of neuropeptide Y which shows selectivity for the prejunctional or neuropeptide Y Y2 receptor. In anaesthetised rats N-acetyl [Leu28,Leu31]neuropeptide Y-(24-36) attenuates cardiac vagal action (a prejunctional or
M J McCloskey et al.
Neuropeptides, 31(2), 193-197 (1997-04-01)
Neuropeptide Y (NPY), a sympathetic cotransmitter, has both prejunctional and postjunctional actions in the cardiovascular system. In anaesthetized rats, the bioassay system used here, NPY attenuates cardiac vagal action (a prejunctional or Y2 action) and increases blood pressure (a postjunctional

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service