Skip to Content
Merck
All Photos(1)

Documents

N0251

Sigma-Aldrich

2-(1-Naphthyl)-5-phenyloxazole

suitable for liquid scintillation spectrometry, crystalline

Synonym(s):

α-NPO

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H13NO
CAS Number:
Molecular Weight:
271.31
Beilstein:
210666
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

form

crystalline

color

light green

suitability

suitable for liquid scintillation spectrometry

storage temp.

−20°C

SMILES string

c1ccc(cc1)-c2cnc(o2)-c3cccc4ccccc34

InChI

1S/C19H13NO/c1-2-8-15(9-3-1)18-13-20-19(21-18)17-12-6-10-14-7-4-5-11-16(14)17/h1-13H

InChI key

WWVFJJKBBZXWFV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service