Skip to Content
Merck
All Photos(4)

Documents

MAK370

Sigma-Aldrich

L-Arginine Assay Kit (Colorimetric)

sufficient for 100 colorimetric tests

Synonym(s):

L-Arginine Quantification Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
NACRES:
NA.84

usage

sufficient for 100 colorimetric tests

application(s)

cosmetics
food and beverages

detection method

colorimetric

relevant disease(s)

cardiovascular diseases

storage temp.

−20°C

General description

L-Arginine (Arg) is a proteogenic, semi-essential amino acid: healthy humans can synthesize L-Arginine using L-Glutamine as a building block; however, premature infants are unable to produce Arg and additional supplementation is required for proper growth and development. Arginine plays pivotal roles in biochemical pathways such as the urea cycle and the biosynthesis of nitric oxide. Arginine and Ammonia concentrations are elevated in patients having a mutation in their ARG1 genes. The mutation causes lower arginase activities – a condition that is known as Argininemia. Arginine has also been advertised as a supplement due to its role in the synthesis of nitric oxide, which helps in vasodilation processes.

Features and Benefits

Compatible with high-throughput handling systems.

Suitability

Suitable for the measurement of L-arginine in biological samples (serum, etc) and beverages (orange juice, etc), the analysis of the relationship of L-arginine intake and nitric oxide production and the analysis of the urea cycle.

Principle

The L-Arginine Assay Kit provides a quick, specific, and easy method for the measurement of total L-arginine concentrations in a wide variety of samples. In this enzyme-based assay, L-arginine is converted into a series of intermediates, which will further react with a probe producing a stable colorimetric signal at A450 nm. The kit is simple to use, sensitive and high-throughput adaptable and can detect as low as 1 nmol/well of L-arginine in biological samples.

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1 - Met. Corr. 1 - Repr. 1B - Resp. Sens. 1 - Skin Corr. 1A

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service