Skip to Content
Merck
All Photos(2)

Documents

MAK070

Sigma-Aldrich

Oxaloacetate Assay Kit

sufficient for 100 colorimetric or fluorometric tests

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
NACRES:
NA.84

usage

sufficient for 100 colorimetric or fluorometric tests

detection method

colorimetric
fluorometric

relevant disease(s)

cancer

storage temp.

−20°C

General description

Oxaloacetate (OAA) is an intermediate of both the TCA cycle and gluconeogenesis. It precedes citrate which is formed by the citrate synthase-mediated condensation of acetyl-CoA and OAA. OAA is also formed by the deamidation of aspartate or condensation of CO2 with pyruvate or PEP. Since mammals do not possess the enzymatic machinery to form TCA cycle intermediates from acetyl-CoA, OAA is one of the anaplerotic entry points via pyruvate and pyruvate carboxykinase. The Oxaloacetate Assay Kit provides a simple, sensitive and rapid means of quantifying OAA in a variety of samples.

Application

Oxaloacetate Assay Kit has been used to determine the concentration of oxaloacetate in samples.

Suitability

Suitable for animal tissues, cell culture (adherent or suspension cells), serum and saliva.

Principle

In this kit, oxaloacetate concentration is determined by a coupled enzyme assay, which results in a colorimetric (570 nm)/fluorometric (lex = 535/lem = 587 nm) product, proportional to the oxaloacetate present. Typical detection ranges for this kit are 2–10 nmole (colorimetric) and 0.2–1 nmole (fluorometric).

replaced by

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ke Tang et al.
Oncogene, 38(44), 6970-6984 (2019-08-15)
Clinical applications of antiangiogenic agents profoundly affect tumor cell behaviors via the resultant hypoxia. To date, how the hypoxia regulates tumor cells remains unclear. Here, we show that hypoxia promotes the growth of human breast tumorigenic cells that repopulate tumors
Zheng Li et al.
Stem cells (Dayton, Ohio), 37(12), 1542-1555 (2019-10-02)
Mitochondrial phosphoenolpyruvate carboxykinase (PCK2) is a rate-limiting enzyme that plays critical roles in multiple physiological processes. The decompensation of PCK2 leads to various energy metabolic disorders. However, little is known regarding the effects of PCK2 on osteogenesis by human mesenchymal
Huabo Wang et al.
The Journal of biological chemistry, 294(14), 5466-5486 (2019-02-14)
Eukaryotic cell metabolism consists of processes that generate available energy, such as glycolysis, the tricarboxylic acid (TCA) cycle, and oxidative phosphorylation (Oxphos), and those that consume it, including macromolecular synthesis, the maintenance of ionic gradients, and cellular detoxification. By converting
The transcription factor Cabut coordinates energy metabolism and the circadian clock in response to sugar sensing.
Bartok O, Teesalu M, Ashwall-Fluss R, et al.
The Embo Journal, 34(11), 1538-1553 (2015)
Qiao Xue et al.
Journal of virology, 96(4), e0191921-e0191921 (2021-12-16)
African swine fever is one of the most serious viral diseases caused by African swine fever virus (ASFV). The metabolic changes induced by ASFV infection remain unknown. Here, porcine alveolar macrophages (PAMs) infected with ASFV was analyzed by ultrahigh-performance liquid

Articles

Sigma article discusses tumor cell metabolic pathways, focusing on aerobic glycolysis and mitochondrial activity.

Fatty acid synthesis supports cancer cell proliferation, essential for membrane generation, protein modification, and bioenergetics.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service