Skip to Content
Merck
All Photos(1)

Documents

I145

Sigma-Aldrich

I-AB-MECA

solid

Synonym(s):

1-[6-[[(4-Amino-3-iodophenyl)methyl]amino]-9H-purin-9-yl]-1-deoxy-N-methyl-β-D-ribofuranuronamide, 3-Iodo-4-aminobenzyl-5′-N-methylcarboxamidoadenosine, N6-(4-Amino-3-iodobenzyl)-5′-N-methylcarbamoyladenosine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H20IN7O4
CAS Number:
Molecular Weight:
525.30
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

form

solid

color

off-white

solubility

45% (w/v) aq 2-hydroxypropyl-β-cyclodextrin: 0.2 mg/mL
DMSO: 10 mg/mL
0.1 M NaOH: 2 mg/mL
0.1 M HCl: 7 mg/mL
H2O: insoluble

storage temp.

−20°C

SMILES string

CNC(=O)[C@H]1O[C@H]([C@H](O)[C@@H]1O)n2cnc3c(NCc4ccc(N)c(I)c4)ncnc23

InChI

1S/C18H20IN7O4/c1-21-17(29)14-12(27)13(28)18(30-14)26-7-25-11-15(23-6-24-16(11)26)22-5-8-2-3-10(20)9(19)4-8/h2-4,6-7,12-14,18,27-28H,5,20H2,1H3,(H,21,29)(H,22,23,24)/t12-,13+,14-,18+/m0/s1

InChI key

LOGOEBMHHXYBID-MOROJQBDSA-N

Gene Information

Biochem/physiol Actions

Reference standard for radioiodinated I-AB-MECA, a widely used, high affinity radioligand for the A3 adenosine receptor.

Legal Information

Sold under license from the National Institutes of Health.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D K Von Lubitz et al.
European journal of pharmacology, 275(1), 23-29 (1995-02-24)
We have previously shown that acute preischemic adenosine A3 receptor stimulation results in an increased postischemic damage, while chronic stimulation of this receptor diminishes it. Since several pathophysiological phenomena are common for both ischemia and seizures, we have explored the
M E Olah et al.
Molecular pharmacology, 45(5), 978-982 (1994-05-01)
The rat A3 adenosine receptor (AR) is a recently characterized AR subtype cloned from testis and brain cDNA libraries. N6-2-(4-Amino-3-[125I]iodophenyl)ethyladenosine, a high affinity A1AR agonist, has served as the only radioligand available for study of the A3AR. The relatively low
K A Jacobson et al.
FEBS letters, 336(1), 57-60 (1993-12-20)
The behavioral effects of a selective A3 adenosine receptor agonist 3-IB-MECA (N6-(3-iodobenzyl)-5'-N-methylcarboxamidoadenosine) in mice and the localization of radioligand binding sites in mouse brain were examined. Low levels of A3 adenosine receptors were detected in various regions of the mouse

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service