Skip to Content
Merck
All Photos(1)

Documents

HYSHP020

Sigma-Aldrich

HyStem®-HP Cell Culture Scaffold Kit

For 7.5 mL of hydrogel scaffold solution

Synonym(s):

Cell Culture Scaffold, HP Cell Culture Kit, HyStem-HP Scaffold Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
NACRES:
NA.75

Quality Level

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

The HyStem®-HP Cell Culture Scaffold Kit provides an excellent starting point for optimizing the matrix for stem cell culture, where slowly released growth factors are crucial in re-creating a stem cell niche. To affect specific cell performance, growth factors or ECM proteins may be added to the HyStem-HP hydrogel. Unlike animal-derived ECM products, this kit contains three fully chemically defined components, which are nonimmunogenic:
  • HyStem®-HP – a thiol-modified hyaluronan (a major constituent of native ECM), carboxymethyl hyaluronic acid-thiopropanoyl hydrazide (CMHA-S, CMHA-DTPH, carboxymethyl hyaluronic acid-DTPH) with heparinthiopropanoyl hydrazide (HP-DTPH). HyStem-HP (CMHA-DTPH with HP-DTPH) contains 99.7 wt% CMHA-DTPH and 0.3 wt% HP-DTPH.
  • Gelin-S – a thiol-modified gelatin (denatured collagen), carboxymethyl gelatin-thiopropanoyl hydrazide (GTN-DTPH, carboxymethyl gelatin-DTPH)
  • Extralink® – a thiol-reactive crosslinker, polyethylene glycol diacrylate (MW = 3,400 g/mole, PEGDA)

Features and Benefits

Customizable, well characterized and consistent, Hystem-HP hydrogel cell culture scaffolds provide a complex, three dimensional environment in which cells are able to proliferate, much as they would in vivo.
  • Customizable - each distinct cell type in a multicellular organism has its own unique microenvironment. Hystem-HP offers the flexibility to tailor the environments of cultured cells to mimic their in vivo counterparts and optimize proliferation and differentiation.
  • Synthetic - As a synthetic matrix, Hystem-HP offers greater control of the cellular environment than ambiguous extracted ECM alternatives. All components are well characterized and consistently formulated. Hystem-HP contains Hyaluronic acid (synthetic), Extralink (synthetic), Gelin-S (porcine), Heparin and Water.
  • Biologically accurate - Rich in hyaluronic acid and denatured collagen, the Hystem-HP hydrogel closely mimics the complex three dimensional cellular environments found in life, providing an optimal environment for culturing your cells.
Hystem-HP enables ionic binding and slow release of growth factors through thiolated heparin; growth factors can be integrated via the matrix rather than cell culture medium. Hystem-HP also enables the attachment of a wide variety of cell types through Gelin-S (thiolated gelatin).

With Hystem-HP, researchers can tailor the incorporation of ECM proteins and growth factors as well as the elasticity of the hydrogel and cell incorporation (encapsulations or top plating) to create the specific 3D environment required by their cells.

Hystem-HP is recommended for both in vivo and in vitro experimentation.

Legal Information

Extralink is a registered trademark of Glycosan BioSystems, Inc.
Gelin is a trademark of Biopharm Ltd.
Hystem is a registered trademark of BIOTIME, INC

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Guangyue Li et al.
Oxidative medicine and cellular longevity, 2022, 3319056-3319056 (2022-10-21)
Advanced glycation end products (AGEs) accumulate in the bone tissue of patients with diabetes mellitus, resulting in oxidative stress, poor bone healing, or regeneration. Irisin, a novel exercise-induced myokine, is involved in the regulation of bone metabolism. However, the effects

Articles

Hystem 3D hydrogels are based on hyaluronic acid, a major component of the extracellular matrix. 3D cell culture can be done using Hystem hydrogels.

Heparin is distinct from heparan sulfate proteoglycans (HS) which is primarily produced by mast cells. While, HS is produced by all cell types. Know more about the difference between heparin and heparin sulfate proteoglycan and select the right component for your study.

Explore the role of hyaluronan, hyaluronic acid structure, hyaluronan synthesis & degradation, and more. Find GAGs, hydrogels, and scaffold kits.

3D cell culture overview. Learn about 2D vs 3D cell culture, advantages of 3D cell culture, and techniques available to develop 3D cell models

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service