Skip to Content
Merck
All Photos(2)

Documents

HPA007045

Sigma-Aldrich

Anti-GNE antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Bifunctional UDP-N-acetylglucosamine 2-epimerase/N-acetylmannosamine kinase antibody produced in rabbit, Anti-UDP-GlcNAc-2-epimerase antibody produced in rabbit, Anti-UDP-GlcNAc-2-epimerase/ManAc kinase) [Includes: UDP-N-acetylglucosamine 2-epimerase antibody produced in rabbit, Anti-Uridine diphosphate-N-acetylglucosamine-2-epimerase antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

DDFDINTRLHTIVRGEDEAAMVESVGLALVKLPDVLNRLKPDIMIVHGDRFDALALATSAALMNIRILHIEGGEVSGTIDDSIRHAITKLAHYHVCCTRSAEQHLISMCEDHDRILLAGCPSYDKLLSAKNKD

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GNE(10020)

Immunogen

Bifunctional UDP-N-acetylglucosamine 2-epimerase/N-acetylmannosamine kinase recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

GNE (Glucosamine (UDP-N-acetyl)-2-epimerase/N-acetylmannosamine kinase) encodes a rate-limiting, bifunctional single protein, UDP-N-acetylglucosamine 2-epimerase and N-acetylmannosamine kinase that is involved in the biosynthesis of sialic acid. Sialic acid is an essential cell surface molecule for several biological processes, including cell adhesion and signal transduction. Mutation in GNE causes distal myopathy with rimmed vacuoles.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71395

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anna Cho et al.
Journal of neurology, neurosurgery, and psychiatry, 85(8), 914-917 (2013-09-13)
GNE myopathy (also called distal myopathy with rimmed vacuoles or hereditary inclusion body myopathy) is an autosomal recessive myopathy characterised by skeletal muscle atrophy and weakness that preferentially involve the distal muscles. It is caused by mutations in the gene
Ichizo Nishino et al.
Journal of neurology, neurosurgery, and psychiatry, 86(4), 385-392 (2014-07-09)
GNE myopathy is an autosomal recessive muscle disease caused by biallelic mutations in GNE, a gene encoding for a single protein with key enzymatic activities, UDP-N-acetylglucosamine 2-epimerase and N-acetylmannosamine kinase, in sialic acid biosynthetic pathway. The diagnosis should be considered
O T Keppler et al.
Science (New York, N.Y.), 284(5418), 1372-1376 (1999-05-21)
Modification of cell surface molecules with sialic acid is crucial for their function in many biological processes, including cell adhesion and signal transduction. Uridine diphosphate-N-acetylglucosamine 2-epimerase (UDP-GlcNAc 2-epimerase) is an enzyme that catalyzes an early, rate-limiting step in the sialic

Articles

Understand sialic acid structure, function, signaling, and modifications. Easily find products for sialic acid research.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service