Skip to Content
Merck
All Photos(1)

Documents

H4025

Sigma-Aldrich

15α-Hydroxytestosterone

Synonym(s):

4-Androstene-15α,17β-diol-3-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H28O3
CAS Number:
Molecular Weight:
304.42
MDL number:
UNSPSC Code:
12352200

drug control

USDEA Schedule III; Home Office Schedule 4.2; regulated under CDSA - not available from Sigma-Aldrich Canada

storage temp.

2-8°C

SMILES string

C[C@]12CC[C@H]3[C@@H](CCC4=CC(=O)CC[C@]34C)[C@@H]1[C@@H](O)C[C@@H]2O

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N Harada et al.
The Journal of biological chemistry, 259(2), 1265-1271 (1984-01-25)
Testosterone 15 alpha-hydroxylase (cytochrome P-450(15) alpha) was purified from female 129/J mouse liver microsomes based on its specific activities in the eluates from the columns of octylamino-Sepharose 4B, hydroxylapatite, DEAE-Bio-Gel A, and CM52 chromatography. The 15 alpha-hydroxylation activity was five
Bradley A Young et al.
General and comparative endocrinology, 136(2), 276-281 (2004-03-19)
The sea lamprey (Petromyzon marinus L.) represents one of the two most ancient classes of vertebrates and possesses a functional hypothalamus-pituitary-gonadal axis. However, the presence and functionality of androgens in the sea lamprey remain elusive. Recently, 15alpha-hydroxytestosterone (15alpha-T) has been
G Smith et al.
The Biochemical journal, 331 ( Pt 3), 783-792 (1998-04-30)
Cytochrome P-450 CYP2D6, human debrisoquine hydroxylase, metabolizes more than 30 prescribed drugs, the vast majority of which are small molecules containing a basic nitrogen atom. In contrast, the similar mouse protein Cyp2d-9 was first characterized as a testosterone 16alpha-hydroxylase. No
Bradley A Young et al.
General and comparative endocrinology, 151(1), 108-115 (2007-02-03)
The sea lamprey (Petromyzon marinus) is one of the earliest extant vertebrates for which the hypothalamic-pituitary-gonadal (HPG) axis has been shown to control and regulate reproduction in a similar fashion to gnathostome vertebrates. While the two forms of gonadotropin-releasing hormones
Formation of 15 alpha-hydroxylated androgens by the testis and other tissues of the sea lamprey, Petromyzon marinus, in vitro.
D E Kime et al.
General and comparative endocrinology, 46(2), 267-270 (1982-02-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service