Skip to Content
Merck
All Photos(1)

Documents

G6251

Sigma-Aldrich

β-Glycerophosphate disodium salt hydrate

≤0.20% L-α-isomer

Synonym(s):

BGP, Glycerol 2-phosphate disodium salt hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(HOCH2)2CHOP(O)(ONa)2 · xH2O
CAS Number:
Molecular Weight:
216.04 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

impurities

≤0.20% L-α-isomer

mp

102-104 °C (lit.)

SMILES string

O.[Na+].[Na+].OCC(CO)OP([O-])([O-])=O

InChI

1S/C3H9O6P.2Na.H2O/c4-1-3(2-5)9-10(6,7)8;;;/h3-5H,1-2H2,(H2,6,7,8);;;1H2/q;2*+1;/p-2

InChI key

ROPZSVKNEIIIDE-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

β-Glycerophosphate is a classical serine-threonine phosphatase inhibitor used in kinase reaction buffers. BGP is often used in combination with other phosphatase/protease inhibitors for broad spectrum inhibition. It functions as an organic phosphate donor and has been used in culture media for mesenchymal stem cell differentiation to osteoblast-type cells. BGP is also used to buffer M17 media for Lactococcus culture in recombinant protein expression.
Initiates differentiation of bone-marrow-derived stem cells embedded in a natural collagen/chitosan gel designed for tissue engineering.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Buffer B70
Cold Spring Harbor Protocols, 2009, 11970-11970 (2009)
Limin Wang et al.
Biomaterials, 31(14), 3976-3985 (2010-02-23)
Chitosan and collagen type I are naturally derived materials used as cell carriers because of their ability to mimic the extracellular environment and direct cell function. In this study beta-glycerophosphate (beta-GP), an osteogenic medium supplement and a weak base, was
Buffer B70 (RNase-free)
Cold Spring Harbor Protocols, 2009, 11977-11977 (2009)
Lucas R Brun et al.
SpringerPlus, 3, 676-676 (2014-12-19)
The aim of this study was to evaluate the effect of strontium ranelate (SrR) on bone mineral density (BMD) and bone turnover markers after 1 year of treatment. Additionally, the effect of SrR in bisphosphonate-naïve patients (BP-naïve) compared to patients previously
Ya-Ting Deng et al.
PloS one, 10(8), e0136572-e0136572 (2015-09-01)
Echinocystic acid (EA) is a natural triterpone enriched in various herbs and has been used for medicinal purposes in China. In the present study, we systematically examined the effects of EA on ovariectomy-induced osteoporosis in rats for the first time.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service