Skip to Content
Merck
All Photos(1)

Key Documents

EHU156191

Sigma-Aldrich

MISSION® esiRNA

targeting human TM4SF1

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
R$1,765.00
50 μG
R$3,150.00

R$1,765.00


Estimated to ship onApril 22, 2025



Select a Size

Change View
20 μG
R$1,765.00
50 μG
R$3,150.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

R$1,765.00


Estimated to ship onApril 22, 2025


description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CATGAAAACTGTGGCAAACGATGTGCGATGCTTTCTTCTGTATTGGCTGCTCTCATTGGAATTGCAGGATCTGGCTACTGTGTCATTGTGGCAGCCCTTGGCTTAGCAGAAGGACCACTATGTCTTGATTCCCTCGGCCAGTGGAACTACACCTTTGCCAGCACTGAGGGCCAGTACCTTCTGGATACCTCCACATGGTCCGAGTGCACTGAACCCAAGCACATTGTGGAATGGAATGTATCTCTGTTTTCTATCCTCTTGGCTCTTGGTGGAATTGAATTCATCTTGTGTCTTATTCAAGTAATAAATGGAGTGCTTGGAGGCATATGTGGCTTTTGCTGCTCTCACCAACAGCAATATGACTGCTAAAAGAACCAACCCAGGACAGAGCCACAATCTTCC

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yu-Cheng Su et al.
mAbs, 6(4), 1069-1083 (2014-05-31)
Modification of antibody class and binding properties typically requires cloning of antibody genes, antibody library construction, phage or yeast display and recombinant antibody expression. Here, we describe an alternative "cloning-free" approach to generate antibodies with altered antigen-binding and heavy chain
Young Ran Park et al.
International journal of oncology, 48(5), 2135-2143 (2016-03-18)
Transmembrane-4-L6 family 1 (TM4SF1) is upregulated in colorectal carcinoma (CRC). However, the mechanism leading to inhibition of the TM4SF1 is not known. In the present study, we investigated the regulation of TM4SF1 and function of microRNAs (miRNAs) in CRC invasion
Dong Xu et al.
Cancer biology & therapy, 21(4), 354-363 (2020-01-08)
Background: Transmembrane-4-L-six-family-1 (TM4SF1) functions to regulate cell growth and mobility and TM4SF1 expression was upregulated in pancreatic cancer. This study further investigated the role of TM4SF1 in regulating pancreatic cancer epithelial-mesenchymal transition (EMT) and angiogenesis and the underlying molecular events.Methods:
Yonghoon Kwon et al.
PloS one, 9(9), e108771-e108771 (2014-09-25)
5' AMP-activated protein kinase (AMPK) is a highly conserved serine-threonine kinase that regulates energy expenditure by activating catabolic metabolism and suppressing anabolic pathways to increase cellular energy levels. Therefore AMPK activators are considered to be drug targets for treatment of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service