Skip to Content
Merck
All Photos(1)

Documents

EHU077661

Sigma-Aldrich

MISSION® esiRNA

targeting human SPP1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

AGCTGGATGACCAGAGTGCTGAAACCCACAGCCACAAGCAGTCCAGATTATATAAGCGGAAAGCCAATGATGAGAGCAATGAGCATTCCGATGTGATTGATAGTCAGGAACTTTCCAAAGTCAGCCGTGAATTCCACAGCCATGAATTTCACAGCCATGAAGATATGCTGGTTGTAGACCCCAAAAGTAAGGAAGAAGATAAACACCTGAAATTTCGTATTTCTCATGAATTAGATAGTGCATCTTCTGAGGTCAATTAAAAGGAGAAAAAATACAATTTCTCACTTTGCATTTAGTCAAAAGAAAAAATGCTTTATAGCAAAATGAAAGAGAACATGAAATGCTTCTTTCTCAGTTTATTGGTTGAATGTGTATCTATTTGAGTCTGGAAATAACTAATGTGTTTGATAATTAGTTTAGTTTGTGGCTTCATGGAAACTCC

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yao Fan et al.
Bone research, 8, 9-9 (2020-03-05)
Osteocytes are mechanosensitive bone cells, but little is known about their effects on tumor cells in response to mechanical stimulation. We treated breast cancer cells with osteocyte-derived conditioned medium (CM) and fluid flow-treated conditioned medium (FFCM) with 0.25 Pa and 1 Pa
Beibei Zhang et al.
Experimental and therapeutic medicine, 20(4), 3633-3642 (2020-08-29)
The metastatic behavior of hepatocellular carcinoma (HCC) is one of the key factors that leads to poor prognosis. The aim of the current study was to determine the changes in metastasis and the proliferation potential of bone marrow mesenchymal stem
Huan Yu et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 35(3), e21405-e21405 (2021-02-10)
Microglia activation and release of pro-inflammatory cytokines have been closely linked to glaucoma. However, the mechanisms that initiate these pathways remain unclear. Here, we investigated the role of a pro-inflammatory cytokine--osteopontin (OPN), in retinal microglia activation process along with the
Sheng-Li Hu et al.
Molecular neurobiology, 52(1), 236-243 (2014-08-26)
Neurosurgical operations may result in surgical injury which would lead to postoperative neurological deficits. Hyperbaric oxygen preconditioning (HBO-PC) may be beneficial for such people. However, the exact mechanism underlying HBO-PC is not well known yet. The aim of this study
Iman A Mohamed et al.
PloS one, 10(4), e0123318-e0123318 (2015-04-18)
Enhanced expression and activity of the Na+/H+ exchanger isoform 1 (NHE1) has been implicated in cardiomyocyte hypertrophy in various experimental models. The upregulation of NHE1 was correlated with an increase in osteopontin (OPN) expression in models of cardiac hypertrophy (CH)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service