Skip to Content
Merck
All Photos(1)

Documents

EHU037471

Sigma-Aldrich

MISSION® esiRNA

targeting human ECT2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

TTTGGATTCTCCGGAATTTGAAAATGTATTTGTAGTCACGGACTTTCAGGATTCTGTCTTTAATGACCTCTACAAGGCTGATTGTAGAGTTATTGGACCACCAGTTGTATTAAATTGTTCACAAAAAGGAGAGCCTTTGCCATTTTCATGTCGCCCGTTGTATTGTACAAGTATGATGAATCTAGTACTATGCTTTACTGGATTTAGGAAAAAAGAAGAACTAGTCAGGTTGGTGACATTGGTCCATCACATGGGTGGAGTTATTCGAAAAGACTTTAATTCAAAAGTTACACATTTGGTGGCAAATTGTACACAAGGAGAAAAATTCAGGGTTGCTGTGAGTCTAGGTACTCCAATTATGAAGCCAGAATGGATTTATAAAGCTTGGGAAAGGCGGAATGAACAGGAT

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zeinab Kosibaty et al.
Laboratory investigation; a journal of technical methods and pathology, 99(4), 551-567 (2018-12-14)
Epithelial cell transforming sequence 2 (ECT2), a guanine nucleotide exchange factor, is predominantly localized in the nucleus of non-transformed cells and functions to regulate cytokinesis. ECT2 is also localized in the cytoplasm of cancer cells. Aberrant cytoplasmic expression of ECT2
Zheng-Qing Fang et al.
Journal of cellular biochemistry, 119(10), 8317-8324 (2018-06-23)
We intended to evaluate miR-490-5p expression in hepatocellular carcinoma (HCC) tissues and detect the potential targets of miR-490-5p. In vitro experiments were conducted to further investigate the biological function of miR-490-5p on HCC cell metastasis. We investigated the abnormally expressed
J Sebastián Gómez-Cavazos et al.
Current biology : CB, 30(16), 3101-3115 (2020-07-04)
Cytokinesis partitions the cell contents to complete mitosis. During cytokinesis, polo-like kinase 1 (PLK1) activates the small GTPase RhoA to assemble a contractile actomyosin ring. PLK1 is proposed to pattern RhoA activation by creating a docking site on the central
Qi Zhang et al.
Theranostics, 10(23), 10769-10790 (2020-09-16)
Rationale: A number of guanine nucleotide exchange factors (GEFs) including epithelial cell transforming factor ECT2 are believed to drive carcinogenesis through activating distinct oncogenic GTPases. Yet, whether GEF-independent activity of ECT2 also plays a role in tumorigenesis remains unclear. Methods:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service