Skip to Content
Merck
All Photos(1)

Key Documents

LBRM TG6

96020937, mouse lymph, Lymphoblast

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
Pricing and availability is not currently available.

Product Name

LBRM TG6, 96020937

biological source

mouse lymph

growth mode

Suspension

karyotype

Not specified

morphology

Lymphoblast

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse Lymphoma Radiation-induced

Cell Line Description

The LBRM TG6 line was derived from the LBRM 33 T cell lymphoma. The cells produce IL-2 in response to IL-1 and PHA. These cells can be used to measure IL-1 activity in a very rapid sensitive assay. Mouse strain: B10.BR.

Culture Medium

IDMEM + 2mM Glutamine + 0.02 mM 2-mercaptoethanol + 10% foetal bovine serum (FBS).

Subculture Routine

Maintain cultures between 200,000 to 1,000,000 cells/ml; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service