Skip to Content
Merck
All Photos(1)

Key Documents

3T3(+3)

89022402, mouse unknown/unspecified, Fibroblast

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

3T3(+3), 89022402

biological source

mouse unknown/unspecified

growth mode

Adherent

karyotype

Not specified

morphology

Fibroblast

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse

Cell Line Description

Please note: Available stocks of this cell line are not contact-inhibited. Contact inhibition is the natural process of arresting cell growth when cells come into contact with each other. It is not possible to be sure how the loss of contact inhibition may affect other characteristics of the cell line. This cell line catalog number continues to be a popular choice.

The key consideration is whether the characteristic of contact inhibition is required for cell line work. For example, it is required when testing the affects of expression of potential oncogenes where the induction of the loss of contact inhibition is being measured, whereas it is of less significance when using the cell line to express recombinant proteins for production and purification.

Culture Medium

DMEM + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:10 i.e. seeding at 1-3x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service