Skip to Content
Merck
All Photos(1)

Key Documents

BS-41601Z

Sigma-Aldrich

GlycoProfile EZGlyco® O-Glycan Prep Kit

new

Chemical O-glycan Release

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352204

conjugate

(O-linked)

technique(s)

HPLC: suitable
mass spectrometry (MS): suitable

Application

  • Useful for release and labeling of O-linked glycans from glycoproteins for Glycomics by Mass Spec, capillary electrophoresis, HPLC or other analysis methods.
  • O-glycan biomarker discovery and screening in biological samples such as mucins, serum and exosomes
  • O-glycan analysis of functional foods such as dairy products

Features and Benefits

O-Glycan sample prep in 5 hours

High recovery

Minimize "peeling"

Easy and safe operation

Legal Information

EZGlyco is a registered trademark of SUMITOMO BAKELITE CO., LTD.
GlycoProfile is a trademark of Sigma-Aldrich Co. LLC

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 3 - Carc. 2 - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B - Skin Sens. 1 - STOT RE 2 - STOT SE 3 - Water-react 2

Target Organs

Blood, Respiratory system

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Learn about O-linked glycan strategies, O-glycosidase actions, how to remove sialic acid residues, β-Elimination, and O-glycan modifications.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service