Skip to Content
Merck
All Photos(1)

Key Documents

AV48984

Sigma-Aldrich

Anti-PCMTD1 antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-FLJ10883, Anti-Protein-L-isoaspartate (D-aspartate) O-methyltransferase domain containing 1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
R$2,017.00

R$2,017.00


Estimated to ship onApril 16, 2025



Select a Size

Change View
100 μL
R$2,017.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

R$2,017.00


Estimated to ship onApril 16, 2025


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

41 kDa

species reactivity

mouse, guinea pig, rat, bovine, dog, human, rabbit, horse

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... PCMTD1(115294)

General description

Mutations in Protein-L-isoaspartate (D-aspartate) O-methyltransferase domain containing 1 (PCMTD1) gene are associated with primary angle closure glaucoma (PACG).[1]

Immunogen

Synthetic peptide directed towards the middle region of human PCMTD1

Application

Anti-PCMTD1 antibody produced in rabbit is suitable for western blotting at a concentration of 2.5μg/ml.

Sequence

Synthetic peptide located within the following region: TGQNTWESKNILAVSFAPLVQPSKNDNGKPDSVGLPPCAVRNLQDLARIY

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Roopam Duvesh et al.
Investigative ophthalmology & visual science, 54(8), 5624-5628 (2013-07-13)
Three loci defined by single nucleotide polymorphisms (SNPs) rs11024102 in PLEKHA7, rs3753841 in COL11A1, and rs1015213 between the PCMTD1 and ST18 genes, recently have been associated with primary angle closure glaucoma (PACG). We explored the genetic association of these SNPs

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service