Skip to Content
Merck
All Photos(2)

Documents

AV44970

Sigma-Aldrich

Anti-CLCC1 antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-Chloride channel CLIC-like 1, Anti-KIAA0761, Anti-MCLC, Anti-RP11-475E11.6

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

61 kDa

species reactivity

dog, human, rabbit, mouse, pig, rat, bovine

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... CLCC1(23155)

Immunogen

Synthetic peptide directed towards the N terminal region of human CLCC1

Application

Anti-CLCC1 antibody produced in rabbit is suitable for western blotting at a concentration of 1.25μg/ml and for immunohistochemistry of paraffin-embedded tissue sections at a concentration of 4-8μg/ml.

Biochem/physiol Actions

Chloride channel CLIC-like 1 (CLCC1; Mid-1 related chloride channel, MCLC) belongs to the family of chloride channels present at the plasma membrane and membranes of intracellular compartments. They are involved in the regulation of cell volume and acidification of intracellular compartments. Chloride channels may modulate cell growth and apoptosis. CLCC1 is expressed in membranes of endoplasmic reticulum, Golgi apparatus, and nucleus.

Sequence

Synthetic peptide located within the following region: MHYDAEIILKRETLLEIQKFLNGEDWKPGALDDALSDILINFKFHDFETW

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xinhua Li et al.
Annual review of physiology, 64, 609-633 (2002-02-05)
Hepatocytes possess chloride channels at the plasma membrane and in multiple intracellular compartments. These channels are required for cell volume regulation and acidification of intracellular organelles. Evidence also supports a role of chloride channels in modulation of apoptosis and cell
M Nagasawa et al.
The Journal of biological chemistry, 276(23), 20413-20418 (2001-03-30)
MID-1 is a Saccharomyces cerevisiae gene encoding a stretch-activated channel. Using MID-1 as a molecular probe, we isolated rat cDNA encoding a protein with four putative transmembrane domains. This gene encoded a protein of 541 amino acids. We also cloned

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service