Skip to Content
Merck
All Photos(1)

Key Documents

SHM03

Sigma-Aldrich

MISSION® ExpressMag® Beads

Increases transduction efficiency

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
R$1,993.00
300 μL
R$4,038.00
1 ML
R$11,184.00
5 ML
R$33,511.00
10 ML
R$63,356.00

R$1,993.00


Estimated to ship onJune 02, 2025


Request a Bulk OrderRequest more information

Select a Size

Change View
100 μL
R$1,993.00
300 μL
R$4,038.00
1 ML
R$11,184.00
5 ML
R$33,511.00
10 ML
R$63,356.00

About This Item

UNSPSC Code:
41106609
NACRES:
NA.51

R$1,993.00


Estimated to ship onJune 02, 2025


Request a Bulk OrderRequest more information

Quality Level

product line

MISSION®

shelf life

3 yr

storage temp.

2-8°C

Related Categories

General description

To view protocols from Sigma collaborators for ExpressMag system visit the link below.
www.sigmaaldrich.com/mission-protocols

Application

When coupled with MISSION® ExpressMag® magnetics, ExpressMag beads will increase the transduction efficiency of lentiviral particles. Increases in transduction efficiency are particularly striking in primary cells and in suspension cells.

Preparation Note

Do NOT FREEZE! Significant decrease in efficacy will occur if ExpressMag bead solution is frozen.

Legal Information

ExpressMag is a registered trademark of Sigma-Aldrich Co. LLC
MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Questions

1–6 of 6 Questions  
  1. What is the typical fold increase in transduction efficiency when using ExpressMag beads?

    1 answer
    1. The transduction efficiency increase observed when using the ExpressMag products will depend on the cell line. It is reasonable to expect a roughly 10-fold or greater increase.

      Helpful?

  2. How does the storage temperature relate to shipping conditions?

    1 answer
    1. The storage conditions that a Sigma-Aldrich catalog and label recommend for products are deliberately conservative. For many products, long-term storage at low temperatures will increase the time during which they are expected to remain in specification and therefore are labeled accordingly. Where short-term storage, shipping time frame, or exposure to conditions other than those recommended for long-term storage will not affect product quality, Sigma-Aldrich will ship at ambient temperature. The products sensitive to short-term exposure to conditions other than their recommended long-term storage are shipped on wet or dry ice. Ambient temperature shipping helps to control shipping costs for our customers. At any time, our customers can request wet- or dry-ice shipment, but the special handling is at customer expense if our product history indicates that the product is stable for regular shipment.

      Helpful?

  3. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

  4. What other viruses have been successfully introduced into cells via the use of ExpressMag beads?

    1 answer
    1. Adenovirus, SIV, Retrovirus, Measles virus and HIV have all been successfully introduced into cells using ExpressMag.

      Helpful?

  5. What MISSION products have been successfully introduced into cells using ExpressMag® beads?

    1 answer
    1. MISSION Lentiviral Particles (various targets/clones), SHC002V, SHC003V and SHC008V have been successfully introduced into cells using ExpressMag beads.

      Helpful?

  6. What cell lines have been transduced successfully using ExpressMag Beads?

    1 answer
    1. The following cell types have been successfully transduced using ExpressMag Beads: PBL, PBMC, T Cells, GMB primary tumor cells, A549, NYGM, T98G, U251, U87,YH-13, YK6-1, KS-1, NIH3T3, K562, Vero, HeLa, CHO, L929, B95a, H9, HUVEC, CD4+ Primary Human T-Cells, Human Bone Marrow Macrophages, Primary Keratinocytes, SKOV3, MM-AN, MEF and CD 34+.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service