Skip to Content
Merck
All Photos(1)

Key Documents

C-27001

Sigma-Aldrich

Osteoblast Growth Medium

Ready-to-use kit including Basal Medium and SupplementMix, 500 ml

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
NACRES:
NA.71

form

aqueous solution

packaging

pkg of 500 ml

technique(s)

cell culture | mammalian: suitable

shipped in

wet ice

General description

Osteoblast Growth Medium (Ready-to-use)

Application

Osteoblast Growth Medium has been optimized for the in vitro cultivation of human osteoblasts. The fetal calf serum in the medium is strictly pretested for optimal performance. The medium is optimized for primary human cells, but can also be used for bovine, murine, and rat osteoblasts as well as osteoblast cell lines.Recommended for:Human Osteoblasts (HOB)

Components

Part of a Cell Culture System offered in a variety of formats:Growth Medium (Ready-to-use): Includes Basal Medium and SupplementMixSupplementMix: Contains all media supplements premixed in one vial

Quality

All lots of PromoCell Osteoblast Growth Medium are subjected to comprehensive quality control tests using primary human osteoblasts. Each lot is checked for growth promoting activity, adherence rate, and typical morphology of the tested osteoblasts. Approved in-house lots of media are used as a reference. In addition, all lots of media have been tested for the absence of microbial contaminants (fungi, bacteria, mycoplasma).

Preparation Note

Final supplement concentrations after addition to the basal medium:Fetal Calf Serum: 0.1 ml / ml

Storage and Stability

Store the Basal Medium at 4 to 8°C in the dark and the SupplementMix at -20°C immediately after arrival. Do not freeze the Basal Medium. If stored properly, the products are stable until the expiry date stated on the label. After adding the SupplementMix to the Basal Medium, the shelf life of the complete medium is 6 weeks at 4 to 8°C. Do not freeze the complete medium. For use, prewarm only an aliquot of the complete medium and keep the remaining medium refrigerated at 4 to 8°C.

Subculture Routine

Click here for more information.

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Primary human osteoblasts and media for the optimal growth and mineralization of human osteoblast cultures. Protocols for handling, cell passaging, media, and product use.

Related Content

Cell culture protocol used to differentiate primary human osteoblasts into bone cells in 2-3 weeks.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service