Skip to Content
Merck
All Photos(1)

Key Documents

41161

Sigma-Aldrich

D-Threono-1,4-lactone

≥95.0% (GC)

Synonym(s):

(3S,4R)-3,4-Dihydroxydihydrofuran-2(3H)-one, D-Threonic acid γ-lactone, D-Threonolactone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H6O4
CAS Number:
Molecular Weight:
118.09
Beilstein:
81075
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

Assay

≥95.0% (GC)

optical activity

[α]/D -27.0±5.0°, c = 1 in H2O

suitability

conforms to structure for Proton NMR spectrum

SMILES string

O[C@H]1[C@H](O)C(OC1)=O

InChI

1S/C4H6O4/c5-2-1-8-4(7)3(2)6/h2-3,5-6H,1H2/t2-,3+/m1/s1

InChI key

SGMJBNSHAZVGMC-GBXIJSLDSA-N

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dhouha Darghouth et al.
Blood, 117(6), e57-e66 (2010-12-08)
Emerging metabolomic tools can now be used to establish metabolic signatures of specialized circulating hematopoietic cells in physiologic or pathologic conditions and in human hematologic diseases. To determine metabolomes of normal and sickle cell erythrocytes, we used an extraction method
Dhouha Darghouth et al.
Haematologica, 96(12), 1861-1865 (2011-08-24)
Overhydrated hereditary stomatocytosis, clinically characterized by hemolytic anemia, is a rare disorder of the erythrocyte membrane permeability to monovalent cations, associated with mutations in the Rh-associated glycoprotein gene. We assessed the red blood cell metabolome of 4 patients with this
Naheed Riaz et al.
Journal of Asian natural products research, 15(10), 1080-1087 (2013-07-05)
Two new sphingolipids plicatin A [(2S,3S,4R)-2-{[(2R)-2-hydroxyoctdecanoyl]amino}hexaeicosane-1,3,4-triol (1)] and plicatin B [(2S,3S,4R,10E)-2-{[(2R)-2-hydroxyoctdecanoyl]amino}tricont-10-ene-1,3,4-triol (2)], together with 4-hydroxybenzaldehyde, scopoletin, uracil, and dl-threonolactone were isolated from the methanolic extract of the whole plant of Chrozophora plicata. The structures of these compounds were established using
Eduardo Cruz-Rus et al.
Biotechnology journal, 7(9), 1110-1121 (2012-08-15)
The term "vitamin" is used to define a number of organic compounds that have to be obtained from different foods because the organism itself cannot synthesize them in the quantities needed to sustain life. Vitamin C is the common name

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service