Skip to Content
Merck
All Photos(1)

Key Documents

BX16411

Supelco

Buffer pH 10.0 Blue

certified reference material

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
Pricing and availability is not currently available.

grade

certified reference material

Quality Level

CofA

current certificate can be downloaded

packaging

pkg of 500 mL

application(s)

environmental
environmental
food and beverages
general analytical
industrial qc

format

mixture

storage temp.

15-25°C

General description

This certified reference material (CRM) is produced in accordance with ISO 17034 and characterized in accordance with ISO/IEC 17025. This CRM is traceable to SI and higher-order standards from NIST through a continuous chain of comparisons. It is prepared using qualified and calibrated balances that comply with ISO 17025 requirements.

Please visit ISO certificates and Site Quality Self-Assessments to access the current certificates of accreditation.

Download your certificate at http://www.sigma-aldrich.com to view certified values, including uncertainty and expiry date.

Application

The pH 10.0 Blue buffer standard is suitable for:
  • Calibration of pH meters and electrodes
  • Quality control of pH measurements
  • Analytical method development and validation

Features and Benefits

  • Solution that is stable, ready to use, and have long minimumshelf life
  • Available in asealed bottle with convenient packaging to protect against contamination
  • Accompanied witha certificate of analysis (CoA) for an effective audit
  • Precise pHcontrol with this solution, ensuring accuracy in experiments
  • Reliable andreproducible results for consistent outcome

Packaging

500 ml in Plastic bottle

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service