Skip to Content
Merck
All Photos(1)

Key Documents

36174

Supelco

Monuron

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H4NHCON(CH3)2
CAS Number:
Molecular Weight:
198.65
Beilstein:
2097922
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

173-174 °C (lit.)

application(s)

agriculture
cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

SMILES string

CN(C)C(=O)Nc1ccc(Cl)cc1

InChI

1S/C9H11ClN2O/c1-12(2)9(13)11-8-5-3-7(10)4-6-8/h3-6H,1-2H3,(H,11,13)

InChI key

BMLIZLVNXIYGCK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Monuron belongs to the arylurea family of herbicides, which is widely used in agriculture.

Application

Monuron may be used as a reference standard in the determination of monuron in rice and corn using high performance liquid chromatography coupled with fluorescence detection combined with ultraviolet decomposition and post-column derivatization.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A highly sensitive and rapid ELISA for the arylurea herbicides diuron, monuron, and linuron.
Schneider P, et al.
Journal of Agricultural and Food Chemistry, 42(2), 413-422 (1994)
W Chu et al.
Journal of agricultural and food chemistry, 55(14), 5708-5713 (2007-06-15)
The immobilization of cobalt ion on different media to catalyze oxone has been investigated. A probe herbicide, Monuron, was effectively degraded by using Co2+/oxone systems. For Co2+ supported on zeolite, 100% of Monuron could be removed within a 10 min
L C Rai et al.
Biometals : an international journal on the role of metal ions in biology, biochemistry, and medicine, 5(1), 13-16 (1992-01-01)
The impact of 2,4-dinitrophenol and chlorophenyl dimethylurea on ATP content, carbon fixation, O2 evolution, nitrogenase activity and Cr uptake of Anabaena doliolum has been studied. 2,4-Dinitrophenol has been found to be more toxic than chlorophenyldimethylurea for all these processes. However
[Hygienic regulation of the monurone content of soil].
Iu N Talakin et al.
Gigiena i sanitariia, (12)(12), 55-57 (1984-12-01)
Maria Bobu et al.
Chemosphere, 63(10), 1718-1727 (2005-11-18)
The photodegradation of monuron (3-(4-chlorophenyl)-1,1-dimethylurea) in aqueous solutions under simulated solar irradiation has been conducted by different advanced oxidation processes (UV/H(2)O(2), UV/H(2)O(2)/Fe(2+), UV/H(2)O(2)/TiO(2), UV/TiO(2), dark H(2)O(2)/Fe(3+)). The degradation rates were always higher for the homogeneous catalysis in photo-Fenton reactions (UV/H(2)O(2)/Fe(2+))

Protocols

-methylcarbamate 10 μg/mL; Diuron; Propham; Siduron; Methiocarb, analytical standard; Linuron 10 μg/mL; Swep 10 μg/mL; Chlorpropham 10 μg/mL; Barban; Neburon

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service