Skip to Content
Merck
All Photos(1)

Key Documents

24544

Hamilton® syringe, 800 series, cemented needle

805N, volume 50 μL, needle size 22s ga (bevel tip), needle L 51 mm (2 in.)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122001
Pricing and availability is not currently available.

material

stainless steel needle (point style 2)

needle size

22s ga (bevel tip)

description

Microliter

feature

needle type N (cemented)

packaging

pkg of 1 ea

manufacturer/tradename

Hamilton 84856

needle L

51 mm (2 in.)

volume

50 μL

Looking for similar products? Visit Product Comparison Guide

General description

  • Same quality performance as 700 series, with additional benefits
  • The extended metal handle is removable and field repairable
  • The plunger stem resists bending
  • Accurate to within ±1% of the total volume
  • Temperature limit: 50°C for cemented needles, 115°C for removable needle
Hamilton syringe is available in gas-tight type that minimizes the gas exchange in GC analysis. It gives accurate volume delivery. The plunger is mostly unsurpassed.[1]

Application

It has been used in stripe assay to understand the attractive or repulsive activity of protein substrate fibronectin and leucine-rich transmembrane protein-2 (FLRT2) using dissociated Hippocampal neurons.[2]

Legal Information

Hamilton is a registered trademark of Hamilton Co.
Microliter is a trademark of Hamilton Co.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stripe Assay to Study the Attractive or Repulsive Activity of a Protein Substrate Using Dissociated Hippocampal Neurons
Yamagishi, Satoru, et al.
Journal of Visualized Experiments, 112, e54096-e54096 (2016)
In Vitro Fertilization and Embryo Transfer: A Manual of Basic Techniques, 06-Dec, 192-192 (2012)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service