Skip to Content
Merck
All Photos(1)

Documents

05-4560

Sigma-Aldrich

Chromium(III) potassium sulfate dodecahydrate

SAJ first grade, ≥97.0%

Synonym(s):

Chrome alum, Potassium chromium(III) sulfate dodecahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CrK(SO4)2 · 12H2O
CAS Number:
Molecular Weight:
499.40
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

grade

SAJ first grade

Assay

≥97.0%

form

crystalline

reaction suitability

reagent type: catalyst
core: chromium

availability

available only in Japan

mp

89 °C (lit.)

SMILES string

O.O.O.O.O.O.O.O.O.O.O.O.[K+].[Cr+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

InChI

1S/Cr.K.2H2O4S.12H2O/c;;2*1-5(2,3)4;;;;;;;;;;;;/h;;2*(H2,1,2,3,4);12*1H2/q+3;+1;;;;;;;;;;;;;;/p-4

InChI key

ZFVHBEKVAITXHW-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R S Liem et al.
Acta morphologica Neerlando-Scandinavica, 22(3), 233-243 (1984-10-01)
The ultrastructure of endochondral bone was studied using an aqueous solution of chromic potassium sulphate as the decalcifying agent. 0.5 mm thick sections of rat tibiae were fixed in buffered glutaraldehyde, immersed in an aqueous solution of 1% chromic potassium
L A Kowalski et al.
Cancer letters, 103(1), 65-69 (1996-05-15)
Tumor promoters such as phorbol esters, teleocidin and okadaic acid increase the numbers of multilayered, transformed foci produced by BPV DNA-transfected C3H/10T1/2 cells. We questioned whether arsenic and chromium, which are known human carcinogens also enhance transformation of BPV DNA-transfected
W Sandritter et al.
Journal of microscopy, 121(Pt 3), 253-259 (1981-03-01)
A method has been elaborated for the demonstration of DNA in the electron microscope. The method uses glutaraldehyde fixed tissue pieces from which RNA has been removed by incubation with RNase. DNA is stained by gallocyanin-chrome alum in the tissue
Rapid demonstration of nucleic acids using "oxidised" gallocyanin and chromic potassium sulphate: methods and applications.
O A Husain et al.
Journal of clinical pathology, 37(1), 99-101 (1984-01-01)
Venous sclerosants: distant ischaemic disorders.
Prescrire international, 20(117), 155-155 (2011-06-18)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service