Skip to Content
Merck
All Photos(1)

Key Documents

RIE156319119

Rieke Metals, Inc.

3-(1,2,3,4-Tetrahydroquinolin-2-ylmethyl)phenylmagnesium bromide 0.25 M in Tetrahydrofuran

0.25 M in Tetrahydrofuran

Sign Into View Organizational & Contract Pricing


About This Item

Molecular Weight:
326.52
MDL number:

Pricing and availability is not currently available. Contact Technical Service

Iupac Name

bromo[3-(3,4-dihydro-2(1H)-isoquinolinylmethyl)phenyl]magnesium

Inchi Code

1S/C16H16N.BrH.Mg/c1-2-6-14(7-3-1)12-17-11-10-15-8-4-5-9-16(15)13-17;;/h1-2,4-9H,10-13H2;1H;/q;;+1/p-1

InChI key

JDPMMMHWPPZUKY-UHFFFAOYSA-M

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
HPA013397SAB2102984HPA071529
biological source

rabbit

biological source

rabbit

biological source

rabbit

biological source

rabbit

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

product line

Prestige Antibodies® Powered by Atlas Antibodies

product line

Prestige Antibodies® Powered by Atlas Antibodies

product line

-

product line

Prestige Antibodies® Powered by Atlas Antibodies

shipped in

wet ice

shipped in

wet ice

shipped in

wet ice

shipped in

wet ice

Country of Origin

US

Physical Form

Liquid

Product Link

https://riekemetals.com/products-services/catnum/3194

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service