Skip to Content
Merck
All Photos(5)

Key Documents

TZSPLKT10

Millipore

Steritest Sampling Kit - Guide + Needle

ready-to-use

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23151818
NACRES:
NA.31
Pricing and availability is not currently available.

sterility

sterile; ethylene oxide treated

Quality Level

feature

ready-to-use

packaging

10 pak of

application(s)

pharmaceutical

storage temp.

15-25°C

General description

Steritest® Sampling kit is as an accessory designed to be installed on Steritest® Neo Devices for sampling or injecting in the canister with a needle (included).
It can be easily mounted onto any Steritest® Neo canisters whenever needed, to sample or inject without risks for your hands or the canister.

Application

Steritest® Sampling kit (Needle + Guide) is used to
  • Sample an aliquot of media from the canisters during or after incubation during sterility testing.
  • Inject in the canister for positive controls

Legal Information

STERITEST is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service