Skip to Content
Merck
All Photos(2)

Documents

MABT390

Sigma-Aldrich

Anti-Syntaxin-2 Antibody, clone 2F2.1

clone 2F2.1, from mouse

Synonym(s):

Syntaxin-2, Epimorphin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

2F2.1, monoclonal

species reactivity

human

technique(s)

immunocytochemistry: suitable
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... STX2(2054)

General description

The protein named Syntaxin-2 or Epimorphin is a multifunctional membrane protein with dual topology. Its C terminus is membrane anchored. When the N terminus is on the cytosolic face it acts as a t-SNARE involved in intracellular vesicle docking and is called Syntaxin-2. When flipped inside out the N terminus hangs out on the extracellular surface (by some nonclassical secretion pathway) it acts as a versatile morphogen and is called Epimorphin. This membrane protein enjoys the double choice of another form of topological alternatives of being targeted to either apical or basolateral surface of an epithelial cell in a regulated way depending on various contexts. When expressed by mesenchymal cells it can instruct epithelial morphogenesis at epithelial mesenchymal interfaces, and Syntaxin-2 is protein that is essential for epithelial cell and tissue morphogenesis and synaptic function in nervous system communication. Syntaxin-2 interacts with synaptotagmin-6 (SYT6) and synaptotagmin-8 (SYT8) in calcium dependent exocytosis of synaptic vesicles and may mediate calcium regulation of exocytosis in the acrosomal reaction in sperm cells as well.

Immunogen

GST-tagged recombinant protein corresponding to human Syntaxin-2.

Application

Anti-Syntaxin-2 Antibody, clone 2F2.1 is a highly specific mouse monoclonal antibody, that targets Syntaxin 2 & has been tested in western blotting & ICC.
Immunocytochemistry Analysis: A 1:50 dilution from a representative lot detected Syntaxin-2 in SH-SY5Y cells.
Research Category
Cell Structure
Research Sub Category
Cytoskeletal Signaling

Quality

Evaluated by Western Blotting in SH-SY5Y cell lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected Syntaxin-2 in 200 µg of SH-SY5Y cell lysate.

Target description

~ 33 kDa observed

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG2aκ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service