MABS2314
Anti-RHBDF2 Antibody, clone RHF2B 11H7
Synonym(s):
Rhomboid family member 2, Rhomboid veinlet-like protein 6, Rhomboid-related protein, iRhom2, inactive rhomboid protein 2
About This Item
Recommended Products
biological source
rat
Quality Level
antibody form
purified antibody
antibody product type
primary antibodies
clone
RHF2B 11H7, monoclonal
mol wt
observed mol wt ~95 kDa
purified by
using protein G
species reactivity
mouse, human
packaging
antibody small pack of 100
technique(s)
western blot: suitable
isotype
IgG2a
epitope sequence
N-terminal half
Protein ID accession no.
UniProt accession no.
storage temp.
2-8°C
Gene Information
mouse ... Rhbdf2(217344)
Specificity
Immunogen
Application
Evaluated by Western Blotting in MCF-7 cell lysates.
Western Blotting Analysis: A 1:500 dilution of this antibody detected RHBDF2/iRhom2 in MCF-7 cell lysate.
Tested Applications
Western Blotting:
Lipopolysaccharide (LPS; 100 ng/mL; 24 h)-treated wild-type macrophage fractions from mouse bone marrow and RHBDF2 knockouts were probed with Cat. No. MABS2314, Anti-RHBDF2, clone RHF2B 11H7 (1:500 Dilution). Proteins were visualized using a Goat Anti-Rat IgG2a secondary antibody conjugated to HRP and a chemiluminescence detection system. Arrow indicates RHBDF2 (~ 98 kDa). (Data courtesy: Katrin Moschke, Deutsches Zentrum für Neurodegenerative Erkrankungen (DZNE), Munich.
Western Blotting Analysis: A representative lot detected RHBDF2 in Western Blotting applications (Weskamp, G., et al. (2020). J Biol Chem. 295(13):4350-4358).
Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.
Target description
Physical form
Reconstitution
Storage and Stability
Other Notes
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service