Skip to Content
Merck
All Photos(2)

Documents

MABS1281

Sigma-Aldrich

Anti-Itk Antibody, clone 2F12

clone 2F12, from mouse

Synonym(s):

Tyrosine-protein kinase ITK/TSK, IL-2-inducible T-cell kinase, Interleukin-2-inducible T-cell kinase, Kinase EMT, T-cell-specific kinase, Tyrosine-protein kinase Lyk

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

2F12, monoclonal

species reactivity

human

species reactivity (predicted by homology)

mouse (based on 100% sequence homology), rat (based on 100% sequence homology)

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... ITK(3702)

General description

Tyrosine-protein kinase ITK/TSK (EC 2.7.10.2; UniProt Q08881; also known as IL-2-inducible T-cell kinase, Interleukin-2-inducible T-cell kinase, Kinase EMT) (T-cell-specific kinase, Tyrosine-protein kinase Lyk) is encoded by the ITK (also known as EMT, LPFS1, LYK) gene (Gene ID 3702) in human. Interleukin-2-inducible T-cell kinase (Itk) is a TEC family non-receptor tyrosine kinase that plays an important roles in signal transduction in T-cells. Upon cell surface receptor stimulation, Itk is recruited to the membrane by the adaptor molecule LAT (linker for activated T cells), where Itk is transphosphorylated at Tyr511 by Lck, Itk Tyr180 then takes place for full activation. Once activated, Itk transduces cellular signaling by activating phospholipase Cγ1 (PLCγ1), leading to activation of nuclear factor of activated T-cells (NFAT), nuclear factor κB (NFκB), and mitogen-activated protein kinase (MAPK) pathways. Itk-/- mice are resistant to developing ovalbumin-induced asthma and exhibit reduced cytokines production by iNKT cells, as well as reduced Th2 effector cytokines and IL-17A productin by T-cells.

Specificity

Clone 2F12 targets the N-terminal region of human, mouse, and rat Itk.

Immunogen

GST-tagged recombinant human/mouse/rat Itk N-terminal fragment.

Application

Detect ITK using this mouse monoclonal Anti-Itk Antibody, clone 2F12, Cat. No. MABS1281, validated for use in Immunoprecipitation and Western Blotting.
Research Category
Signaling
Western Blotting Analysis: A 1:1000 dilution from a representative lot detected Itk in 10 µg of HuT-78 human T-cell lymphoma cell lysate.

Immunoprecipitation Analysis: A representative lot immunoprecipitated Itk from both parental and Lck-negative (J.CaM1) Jurkat cells. Itk immunoprecipitated from the parental Jurkat cells was found with increased tyrosine phosphorylation following TCR cross-linking or pervanadate treatment, while only very weak tyrosine phosphorylation was seen with Itk from J.CaM1 upon prolonged pervanadate treatment (Heyeck, S.D., et al. (1997). J. Biol. Chem. 272(40):25401-25408).

Immunoprecipitation Analysis: A representative lot immunoprecipitated tyrosine-phosphorylated Itk from Jurkat cells stimulated by cross-linking of endogenous cell surface CD3 or transfected CD8 extracellular domain/CD28 cytoplasmic domain chimera (Heyeck, S.D., et al. (1997). J. Biol. Chem. 272(40):25401-25408).

Western Blotting Analysis: A representative lot detected baculovirus expressed Itk from infected Sf9 insect cells following ltk immunoprecipitation by the same antibody. The overexpressed ltk was found autophosphorylated on tyrosine when expressed alone in Sf9 and increased ltk tyrosine phosphorylation was seen when co-expressed with Lck (Heyeck, S.D., et al. (1997). J. Biol. Chem. 272(40):25401-25408).

Quality

Evaluated by Western Blotting in Jurkat cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected Itk in 10 µg of Jurkat cell lysate.

Target description

~71 kDa observed. 71.83/72.29/72.49 (human/mouse/rat) kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified.
Purified mouse IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tone Berge et al.
PloS one, 5(3), e9761-e9761 (2010-03-23)
The chemokine CXCL12/SDF-1alpha interacts with its G-protein coupled receptor CXCR4 to induce migration of lymphoid and endothelial cells. T cell specific adapter protein (TSAd) has been found to promote migration of Jurkat T cells through interaction with the G protein

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service