Skip to Content
Merck
All Photos(3)

Documents

MABD175

Sigma-Aldrich

Anti-SHH Antibody, clone 5H4

clone 5H4, from mouse

Synonym(s):

HHG-1, Sonic hedgehog protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

5H4, monoclonal

species reactivity

mouse, human, monkey

technique(s)

flow cytometry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG1

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... SHH(6469)

General description

Sonic hedgehog protein (SHH) or alternatively known as HHG-1, is a very important developmental intercellular signaling factor. SHH protein is cleaved into to active forms, an N-terminal product that stays associated with the cell membrane or forms freely diffusible multimers and a C-terminal form that diffuses into the extracellular matrix. SHH is expressed in fetal intestine, liver, lung and kidney but not normally in adult tissues except during some disease state like cancerous tumors. The intercellular signaling triggered by SHH is essential for a variety of patterning events during development including the development of the notochord, neural tube and plate, as well as the polarizing of the anterior-posterior axis of developing limb buds. Because of its importance in patterning during development, defects in SHH are the cause of a number of abnormalities including structural eye and brain abnormalities, teeth and bone malformations and finally limb and bone developmental defects. EMD-Millipore’s Anti-SHH mouse monoclonal antibody has been tested in western blot against recombinant antigen and cell lysates from HepG2, PANC-1, HeLa, SK-N-SH, F9, NIH3T3 and COS7 cells and in paraffin embedded immunohistochemistry of human liver cancer tissue sections. The antibody has also been successfully tested by flow cytometry against HeLa cells in culture.

Immunogen

Purified recombinant fragment of human SHH expressed in E. Coli.

Application

Anti-SHH Antibody, clone 5H4 is a highly specific mouse monoclonal antibody, that targets Sonic Hedgehog & has been tested in western blotting, Flow Cytometry & IHC.
Immunohistochemistry Analysis: A 1:200-1,000 dilution from a representative lot detected SHH in liver cancer tissue.

Flow Cytometry Analysis: A 1:200-400 dilution from a representative lot detected SHH in non serum starved HeLa cells.

Optimal working dilutions must be determined by end user.
Research Category
Stem Cell Research
Research Sub Category
Pluripotent & Early Differentiation

Quality

Evaluated by Western Blotting in LNCaP, HepG2, PANC-1, HeLa, SK-N-SH, F9, NIH3T3, COS7 cell lysates.

Western Blotting Analysis: A 1:500-2,000 dilution of this antibody detected SHH in LNCaP, HepG2, PANC-1, HeLa, SK-N-SH, F9, NIH3T3, COS7 cell lysates.

Target description

~50 kDa observed. Uncharacterized bands may appear in some lysate(s).

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal in PBS with up to 0.1% sodium azide and 0.05% protein stabilizer.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
LNCaP, HepG2, PANC-1, HeLa, SK-N-SH, F9, NIH3T3, COS7 cell lysates

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

David Engelhardt et al.
Genesis (New York, N.Y. : 2000), 62(3), e23602-e23602 (2024-05-09)
Cilia play a key role in the regulation of signaling pathways required for embryonic development, including the proper formation of the neural tube, the precursor to the brain and spinal cord. Forward genetic screens were used to generate mouse lines
Carolina Pinzon-Guzman et al.
The Journal of clinical investigation, 130(8), 4396-4410 (2020-05-20)
Esophageal atresia (EA/TEF) is a common congenital abnormality present in 1 of 4000 births. Here we show that atretic esophagi lack Noggin (NOG) expression, resulting in immature esophagus that contains respiratory glands. Moreover, when using mouse esophageal organoid units (EOUs)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service