Skip to Content
Merck
All Photos(1)

Key Documents

DX0030

Supelco

Decane

Synonym(s):

Decane, extraction

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)8CH3
CAS Number:
MDL number:
UNSPSC Code:
12352001
EC Index Number:
204-686-4
Assay:
≥98% (GC)
bp:
174 °C/1013 mbar
vapor pressure:
1 hPa ( 16 °C)
Pricing and availability is not currently available.

vapor pressure

1 hPa ( 16 °C)

Quality Level

Assay

≥98% (GC)

form

liquid

color

APHA: ≤10

bp

174 °C/1013 mbar

mp

-30 °C

transition temp

flash point 51 °C

density

0.73 g/cm3 at 20 °C

shipped in

ambient

storage temp.

room temp

SMILES string

C(CCCCC)CCCC

InChI

1S/C10H22/c1-3-5-7-9-10-8-6-4-2/h3-10H2,1-2H3

InChI key

DIOQZVSQGTUSAI-UHFFFAOYSA-N

Analysis Note

Assay (GC): 98% min
Color (APHA): 10 max
Form: Clear liquid
Infrared Spectrum: Conforms to standard

Pictograms

FlameHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Asp. Tox. 1 - Flam. Liq. 3

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

114.8 °F - closed cup

Flash Point(C)

46.0 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service